Search
Search

Entrez Gene ID: | 55539 |
Gene Name: | KCNQ1 downstream neighbor (non-protein coding) |
Gene Aliases: |
BWRT, HSA404617 |
Location: |
Chr.11:2870033-2872105 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ1DN | NR_024627.1 | 1 | 221 | |
| AB039920.1 | 1 | 221 | ||
| AJ404617.2 | 1 | 191 | CAB95125.2 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759795 | Chr.11:2767814 - 2942382 on Build GRCh38 | Gain |
|
| nsv951292 | Chr.11:2792271 - 2927870 on Build GRCh38 | Deletion |
|
| nsv825715 | Chr.11:2855771 - 2890367 on Build GRCh38 | Gain |
|
| nsv8765 | Chr.11:2861205 - 2871450 on Build GRCh38 | Gain+Loss |
|
| nsv469930 | Chr.11:2831299 - 2924715 on Build GRCh38 | Loss |
|
| dgv1575n54 | Chr.11:2863338 - 2899436 on Build GRCh38 | Loss |
|
| nsv1053066 | Chr.11:2829552 - 2872696 on Build GRCh38 | Gain |
|
| nsv832050 | Chr.11:2868540 - 2963987 on Build GRCh38 | Loss |
|
| nsv467655 | Chr.11:2864498 - 2891716 on Build GRCh38 | Loss |
|
| nsv553111 | Chr.11:2842057 - 2904391 on Build GRCh38 | Loss |
|
| dgv1574n54 | Chr.11:2848935 - 2889391 on Build GRCh38 | Loss |
|