Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02294665_cn
          See other BOC CNV Assays ›
          Gene Symbol
          BOC
          Assay Reference Genome
          Location

          Chr.3:113273284 on build GRCh38
          Cytoband
          3q13.2
          Assay ID Hs02294665_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          91653

          Gene Name:

          BOC cell adhesion associated, oncogene regulated

          Gene Aliases:

          CDON2

          Location:

          Chr.3:113211003-113287463 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 10 - Intron 10
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          BOC NM_001301861.1 NP_001288790.1
          NM_033254.3 NP_150279.1
          XM_005247891.2 XP_005247948.1
          XM_005247892.2 XP_005247949.1
          XM_005247897.2 XP_005247954.1
          XM_011513305.2 XP_011511607.1
          XM_017007441.1 XP_016862930.1
          XM_017007442.1 XP_016862931.1
          XM_017007443.1 XP_016862932.1
          XM_017007444.1 XP_016862933.1
          XM_017007445.1 XP_016862934.1
          XM_017007446.1 XP_016862935.1
          XM_017007447.1 XP_016862936.1
          XM_017007448.1 XP_016862937.1
          XM_017007449.1 XP_016862938.1
          XM_017007450.1 XP_016862939.1
          XM_017007451.1 XP_016862940.1
          XM_017007452.1 XP_016862941.1
          XM_017007453.1 XP_016862942.1
          XM_017007454.1 XP_016862943.1
          AK074556.1 BAC11057.1
          AK090455.1 BAC03436.1
          AY027658.1 AAK14795.1
          AY358328.1 AAQ88694.1
          BC136390.1
          BX647184.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1013506 Chr.3:113171761 - 113962104 on Build GRCh38 Gain CFAP44-AS1 MIR8076 MIR4446 SPICE1 GRAMD1C NAA50 CFAP44 ZDHHC23 SIDT1 ATP6V1A USF3 BOC
          nsv829677 Chr.3:113138228 - 113320140 on Build GRCh38 Gain CFAP44 LOC101929717 BOC
          nsv591293 Chr.3:112266260 - 113293950 on Build GRCh38 Loss NEPRO SLC9C1 LOC101929694 LOC105374042 CCDC80 BTLA SLC35A5 LOC100129297 CD200R1 CD200R1L ATG3 GTPBP8 CFAP44 LINC01279 CD200 LOC107986113 LOC101929717 BOC

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          smoothened signaling pathway
          axon guidance
          positive regulation of myoblast differentiation
          positive regulation of muscle cell differentiation
          mitophagy in response to mitochondrial depolarization
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline