Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02362095_cn
          See other MARK3 CNV Assays ›
          Gene Symbol
          MARK3
          Assay Reference Genome
          Location

          Chr.14:103462426 on build GRCh38
          Cytoband
          Assay ID Hs02362095_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          4140

          Gene Name:

          microtubule affinity regulating kinase 3

          Gene Aliases:

          CTAK1, KP78, PAR1A, Par-1a, VIPB

          Location:

          Chr.14:103385415-103503831 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 9 - Intron 10
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MARK3 NM_001128918.3 NP_001122390.2
          NM_001128919.3 NP_001122391.2
          NM_001128920.3 NP_001122392.2
          NM_001128921.3 NP_001122393.2
          NM_001411055.1 NP_001397984.1
          NM_001411056.1 NP_001397985.1
          NM_001437351.1
          NM_001437366.1
          NM_001437367.1
          NM_001437368.1
          NM_001437369.1
          NM_001438023.1
          NM_002376.7 NP_002367.5
          XM_005267641.6 XP_005267698.2
          XM_005267642.6 XP_005267699.2
          XM_005267643.6 XP_005267700.2
          XM_011536769.4 XP_011535071.1
          XM_017021291.3 XP_016876780.1
          XM_017021292.3 XP_016876781.1
          XM_017021294.3 XP_016876783.1
          XM_017021295.3 XP_016876784.1
          XM_017021297.3 XP_016876786.1
          XM_017021298.3 XP_016876787.1
          XM_017021299.3 XP_016876788.1
          XM_017021301.3 XP_016876790.1
          XM_047431377.1 XP_047287333.1
          XM_047431378.1 XP_047287334.1
          XM_047431379.1 XP_047287335.1
          XM_047431381.1 XP_047287337.1
          XM_047431382.1 XP_047287338.1
          XM_047431383.1 XP_047287339.1
          XM_047431384.1 XP_047287340.1
          XM_047431385.1 XP_047287341.1
          XM_047431386.1 XP_047287342.1
          XM_047431387.1 XP_047287343.1
          XM_047431388.1 XP_047287344.1
          XM_047431389.1 XP_047287345.1
          XM_047431390.1 XP_047287346.1
          XM_047431391.1 XP_047287347.1
          XM_047431392.1 XP_047287348.1
          XM_047431393.1 XP_047287349.1
          XM_047431394.1 XP_047287350.1
          XM_047431395.1 XP_047287351.1
          XM_047431396.1 XP_047287352.1
          AF387637.1 AAK82367.1
          AF465413.1 AAL69982.1
          AK307448.1
          BC024773.1 AAH24773.1
          BX161395.1 CAD61882.1
          M80359.1 AAA59991.1
          U64205.1 AAC15093.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv832883 Chr.14:103461350 - 103654616 on Build GRCh38 Loss TRMT61A MARK3 COA8 CKB BAG5 KLC1
          nsv6510896 Chr.14:103462009 - 103466935 on Build GRCh38 Deletion MARK3
          nsv1054574 Chr.14:103210111 - 105365114 on Build GRCh38 Loss NUDT14 MIR203B C14orf180 CLBA1 CDCA4 TDRD9 LOC105378183 BRF1 LOC124903408 INF2 KIF26A SIVA1 BAG5 KLC1 TMEM179 EIF5 AHNAK2 EIF5-DT LINC02691 PACS2 PLD4 MIR12121 RD3L MIR4710 BTBD6 COA8 KIF26A-DT CKB AKT1 VESTAR LINC02280 MIR6765 TRMT61A SNORA28 MARK3 PPP1R13B-DT ZFYVE21 CEP170B ATP5MJ LOC105370689 ASPG ADSS1 JAG2 GPR132 XRCC3 PPP1R13B LINC02298 MIR203A ZBTB42

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          protein phosphorylation
          regulation of G2/M transition of mitotic cell cycle
          negative regulation of hippo signaling
          intracellular signal transduction
          negative regulation of protein localization to nucleus
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          protein phosphatase binding
          tau protein binding
          tau-protein kinase activity
          protein serine kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline