Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02370700_cn
          See other KCNQ1DN CNV Assays ›
          Gene Symbol
          KCNQ1DN
          Assay Reference Genome
          Location

          Chr.11:2870824 on build GRCh38
          Cytoband
          11p15.4
          Assay ID Hs02370700_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          55539

          Gene Name:

          KCNQ1 downstream neighbor (non-protein coding)

          Gene Aliases:

          BWRT, HSA404617

          Location:

          Chr.11:2870033-2872105 on Build GRCh38

          Assay Gene Location:

          Within Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          KCNQ1DN NR_024627.1 1 792
          AB039920.1 1 792
          AJ404617.2 1 762 CAB95125.2

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2759795 Chr.11:2767814 - 2942382 on Build GRCh38 Gain SLC22A18 CDKN1C SLC22A18AS KCNQ1DN KCNQ1-AS1 PHLDA2 KCNQ1
          nsv951292 Chr.11:2792271 - 2927870 on Build GRCh38 Deletion SLC22A18 CDKN1C SLC22A18AS KCNQ1DN KCNQ1-AS1 KCNQ1
          nsv825715 Chr.11:2855771 - 2890367 on Build GRCh38 Gain CDKN1C SLC22A18AS KCNQ1DN KCNQ1-AS1
          nsv8765 Chr.11:2861205 - 2871450 on Build GRCh38 Gain+Loss KCNQ1DN KCNQ1-AS1
          nsv469930 Chr.11:2831299 - 2924715 on Build GRCh38 Loss SLC22A18 CDKN1C SLC22A18AS KCNQ1DN KCNQ1-AS1 KCNQ1
          dgv1575n54 Chr.11:2863338 - 2899436 on Build GRCh38 Loss CDKN1C SLC22A18AS KCNQ1DN
          nsv1053066 Chr.11:2829552 - 2872696 on Build GRCh38 Gain KCNQ1DN KCNQ1-AS1 KCNQ1
          nsv832050 Chr.11:2868540 - 2963987 on Build GRCh38 Loss SLC22A18 CDKN1C SLC22A18AS NAP1L4 KCNQ1DN PHLDA2 SNORA54
          nsv467655 Chr.11:2864498 - 2891716 on Build GRCh38 Loss CDKN1C SLC22A18AS KCNQ1DN
          nsv553111 Chr.11:2842057 - 2904391 on Build GRCh38 Loss SLC22A18 CDKN1C SLC22A18AS KCNQ1DN KCNQ1-AS1 KCNQ1
          dgv1574n54 Chr.11:2848935 - 2889391 on Build GRCh38 Loss CDKN1C SLC22A18AS KCNQ1DN KCNQ1-AS1 KCNQ1

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline