Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02386806_cn
          See other AHSA2 CNV Assays ›
          Gene Symbol
          AHSA2
          Assay Reference Genome
          Location

          Chr.2:61188476 on build GRCh38
          Cytoband
          2p15
          Assay ID Hs02386806_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          130872

          Gene Name:

          AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)

          Gene Aliases:

          Hch1

          Location:

          Chr.2:61177418-61191210 on Build GRCh38

          Assay Gene Location:

          Within Exon 6
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          AHSA2 NM_001321305.1 6 2666 NP_001308234.1
          XM_011532541.2 8 3018 XP_011530843.1
          XM_011532543.2 9 3302 XP_011530845.1
          XM_011532545.2 8 1726 XP_011530847.1
          XM_017003377.1 7 1624 XP_016858866.1
          XM_017003378.1 9 2977 XP_016858867.1
          AK091998.1 1 2198

          Target Gene Details

          Entrez Gene ID:

          9736

          Gene Name:

          ubiquitin specific peptidase 34

          Gene Aliases:

          -

          Location:

          Chr.2:61187455-61470714 on Build GRCh38

          Assay Gene Location:

          Within Exon 80
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          USP34 NM_014709.3 80 10289 NP_055524.3
          AB011142.2 81 10344 BAA25496.2
          AB018272.1 23 3218 BAA34449.1
          AJ586138.1 78 9814 CAE51938.1
          AK001193.1 20 2880
          AK022637.1 10 1329
          AK023171.1 12 1566
          AL050092.1 15 2385 CAB43264.1
          AL050376.1 1 463
          BC022783.1 17 2588 AAH22783.1
          BC062325.1 8 1043 AAH62325.1
          BC107761.1 7 954 AAI07762.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2760616 Chr.2:61136348 - 61200572 on Build GRCh38 Loss C2orf74 KIAA1841 LOC339803 AHSA2 USP34

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          chaperone cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of ATPase activity
          regulation of transcription, DNA-templated
          ubiquitin-dependent protein catabolic process
          Wnt signaling pathway
          histone deubiquitination
          protein deubiquitination
          protein K48-linked deubiquitination
          positive regulation of canonical Wnt signaling pathway
          ATPase activator activity
          protein binding
          chaperone binding
          cysteine-type endopeptidase activity
          thiol-dependent ubiquitin-specific protease activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline