Search
Search

Entrez Gene ID: | 2185 |
Gene Name: | protein tyrosine kinase 2 beta |
Gene Aliases: |
CADTK, CAKB, FADK2, FAK2, PKB, PTK, PYK2, RAFTK |
Location: |
Chr.8:27311482-27459391 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTK2B | NM_173174.2 | NP_775266.1 | ||
| BC036651.2 | AAH36651.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616717 | Chr.8:27277090 - 27359993 on Build GRCh38 | Gain |
|
| esv3616719 | Chr.8:27304061 - 27359840 on Build GRCh38 | Loss |
|
| dgv1390e214 | Chr.8:27303974 - 27359840 on Build GRCh38 | Gain |
|
Set Membership: |
|