Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02632158_cn
          See other HADHA CNV Assays ›
          Gene Symbol
          HADHA
          Assay Reference Genome
          Location

          Chr.2:26238936 on build GRCh38
          Cytoband
          Assay ID Hs02632158_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          3030

          Gene Name:

          hydroxyacyl-CoA dehydrogenase trifunctional multienzyme complex subunit alpha

          Gene Aliases:

          ECHA, GBP, HADH, LCEH, LCHAD, MLCL AT, MTPA, TP-ALPHA

          Location:

          Chr.2:26190635-26244632 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 3 - Exon 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          HADHA NM_000182.5 NP_000173.2
          AK293398.1 BAG56906.1
          AK299258.1 BAG61288.1
          AK302532.1 BAG63804.1
          AK313027.1 BAG35861.1
          BC009235.2 AAH09235.1
          CN264228.1
          D16480.1 BAA03941.1
          FJ626228.1
          GU727642.1 ADU87643.1
          U04627.1 AAA56664.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6340989 Chr.2:26235255 - 26241536 on Build GRCh38 Deletion HADHA
          esv33316 Chr.2:25440017 - 26672800 on Build GRCh38 Loss ASXL2 HADHA RAB10 DTNB GAREM2 CIB4 KIF3C HADHB DRC1 SELENOI CIMIP2C ADGRF3 OTOF

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid metabolic process
          fatty acid beta-oxidation
          response to insulin
          cardiolipin acyl-chain remodeling
          catalytic activity
          3-hydroxyacyl-CoA dehydrogenase activity
          acetyl-CoA C-acetyltransferase activity
          enoyl-CoA hydratase activity
          protein binding
          oxidoreductase activity
          long-chain-3-hydroxyacyl-CoA dehydrogenase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          transferase activity
          lyase activity
          3-hydroxyacyl-CoA dehydratase activity
          long-chain acyl-CoA hydrolase activity
          NAD+ binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline