Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02715994_cn
          See other RIMKLB CNV Assays ›
          Gene Symbol
          RIMKLB
          Assay Reference Genome
          Location

          Chr.12:8750083 on build GRCh38
          Cytoband
          Assay ID Hs02715994_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          57494

          Gene Name:

          ribosomal modification protein rimK like family member B

          Gene Aliases:

          FAM80B, NAAGS, NAAGS-I

          Location:

          Chr.12:8668638-8783091 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 7 - Intron 7
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          RIMKLB NM_001297776.2 NP_001284705.1
          NM_001352267.2 NP_001339196.1
          NM_001352268.2 NP_001339197.1
          NM_001352269.2 NP_001339198.1
          NM_001352270.2 NP_001339199.1
          NM_001352271.2 NP_001339200.1
          NM_020734.5 NP_065785.2
          NR_123740.2
          NR_147945.2
          NR_147946.2
          NR_147947.2
          NR_147948.2
          NR_147949.2
          NR_147950.2
          XM_017019684.2 XP_016875173.1
          XM_017019686.2 XP_016875175.1
          XM_017019687.2 XP_016875176.1
          XM_024449086.2 XP_024304854.1
          XM_024449087.2 XP_024304855.1
          XM_024449089.2 XP_024304857.1
          XM_024449090.2 XP_024304858.1
          XM_024449091.2 XP_024304859.1
          XM_024449092.2 XP_024304860.1
          XM_024449094.2 XP_024304862.1
          XM_024449095.2 XP_024304863.1
          XM_047429180.1 XP_047285136.1
          XM_047429181.1 XP_047285137.1
          XM_047429182.1 XP_047285138.1
          XM_047429183.1 XP_047285139.1
          XM_047429184.1 XP_047285140.1
          XM_047429185.1 XP_047285141.1
          XM_047429186.1 XP_047285142.1
          XM_047429187.1 XP_047285143.1
          XM_047429188.1 XP_047285144.1
          XM_047429189.1 XP_047285145.1
          XM_047429190.1 XP_047285146.1
          XM_047429191.1 XP_047285147.1
          XM_047429192.1 XP_047285148.1
          XM_047429193.1 XP_047285149.1
          XM_047429194.1 XP_047285150.1
          XM_047429195.1 XP_047285151.1
          XM_047429196.1 XP_047285152.1
          XM_047429197.1 XP_047285153.1
          AB033064.1 BAA86552.2
          AK302847.1 BAH13820.1
          DB265022.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1051530 Chr.12:8442286 - 9951690 on Build GRCh38 Gain PZP AICDA CD69 CLEC2D A2ML1-AS1 M6PR A2M-AS1 CLEC6A KLRG1 DDX12P CLEC4E MIR1244-3 CLEC2B LOC374443 CLEC4D KLRF2 KLRF1 LINC02972 LINC02367 CLEC12A-AS1 CLEC2A LOC728715 CLEC12A A2M RIMKLB LINC00612 PHC1 LOC105369649 LINC00987 LOC642846 LINC02470 KLRB1 LOC408186 MFAP5 CLECL1P A2ML1 LOC105369728 A2MP1 LOC101928030

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          glutamine family amino acid metabolic process
          nucleotide binding
          ATP binding
          ligase activity
          metal ion binding
          N-acetyl-L-aspartate-L-glutamate ligase activity
          citrate-L-glutamate ligase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline