Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02716464_cn
          See other MAPK7 CNV Assays ›
          Gene Symbol
          MAPK7
          Assay Reference Genome
          Location

          Chr.17:19382304 on build GRCh38
          Cytoband
          Assay ID Hs02716464_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5598

          Gene Name:

          mitogen-activated protein kinase 7

          Gene Aliases:

          BMK1, ERK4, ERK5, PRKM7

          Location:

          Chr.17:19377750-19383544 on Build GRCh38

          Assay Gene Location:

          Within Exon 6
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MAPK7 NM_002749.4 5 2139 NP_002740.2
          NM_139032.3 3 1955 NP_620601.1
          NM_139033.3 5 2358 NP_620602.2
          NM_139034.3 5 2051 NP_620603.2
          XM_006721557.4 6 2157 XP_006721620.1
          XM_006721558.4 6 2376 XP_006721621.1
          XM_006721559.4 6 2069 XP_006721622.1
          XM_011523957.4 3 1648 XP_011522259.1
          XM_047436401.1 5 2150 XP_047292357.1
          XM_047436402.1 6 2253 XP_047292358.1
          XM_047436403.1 6 2165 XP_047292359.1
          XM_047436404.1 4 2404 XP_047292360.1
          XM_047436405.1 5 2147 XP_047292361.1
          XM_047436406.1 5 2235 XP_047292362.1
          XM_047436407.1 5 2139 XP_047292363.1
          AB209611.1 1 3776 BAD92848.1
          AK295376.1 5 2081 BAG58335.1
          AY534741.1 3 2108 AAS38577.1
          BC007404.2 3 1952 AAH07404.1
          BC007992.2 3 1952 AAH07992.1
          BC009963.2 5 2108 AAH09963.1
          BC030134.1 5 2023 AAH30134.1
          U25278.1 5 2081 AAA81381.1
          U29725.1 5 2222 AAA82931.1
          U29726.1 5 2055 AAA82932.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4235211 Chr.17:19248750 - 19538890 on Build GRCh38 Duplication MFAP4 LINC02094 EPN2 EPN2-AS1 B9D1 RPS2P46 LOC105371574 RNF112 SLC47A1 MAPK7 MIR1180 EPN2-IT1
          nsv4247486 Chr.17:19371687 - 19382787 on Build GRCh38 Duplication B9D1 MAPK7
          esv3582561 Chr.17:19240831 - 19585268 on Build GRCh38 Loss MFAP4 RPS2P46 LOC105371574 SNORA59B SLC47A1 MAPK7 LINC02094 EPN2 EPN2-AS1 B9D1 RNF112 MIR1180 EPN2-IT1
          nsv1061317 Chr.17:19047542 - 20322438 on Build GRCh38 Gain MFAP4 LOC102724624 GRAP GRAPL-AS1 SNORD3A RPS2P46 CCDC144CP LOC100131943 SPECC1-DT SLC47A1P2 SLC47A2 EPN2-AS1 RNF112 EPN2-IT1 SPECC1 GRAPL SNORD3C SNORD3B-2 LOC105371574 SNORA59B SLC47A1 MAPK7 LINC02094 SNORD3B-1 AKAP10 LOC388436 EPN2 B9D1 ALDH3A1 SNORD3D ALDH3A2 ULK2 MIR1180
          nsv953845 Chr.17:19377788 - 19387787 on Build GRCh38 Deletion MFAP4 B9D1 MAPK7

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          signal transduction
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          cell differentiation
          calcineurin-NFAT signaling cascade
          cellular response to stress
          negative regulation of heterotypic cell-cell adhesion
          negative regulation of smooth muscle cell apoptotic process
          intracellular signal transduction
          negative regulation of apoptotic process
          regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of inflammatory response
          positive regulation of protein metabolic process
          negative regulation of response to cytokine stimulus
          cellular response to hydrogen peroxide
          negative regulation of calcineurin-NFAT signaling cascade
          cellular response to growth factor stimulus
          cellular response to laminar fluid shear stress
          cellular response to transforming growth factor beta stimulus
          intracellular signaling cassette
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          negative regulation of endothelial cell apoptotic process
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase activity
          enzyme inhibitor activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          mitogen-activated protein kinase binding
          protein serine kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline