Search
Search

Entrez Gene ID: | 51095 |
Gene Name: | tRNA nucleotidyl transferase 1 |
Gene Aliases: |
CCA1, CGI-47, MtCCA, RPEM, SIFD |
Location: |
Chr.3:3126940-3153435 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRNT1 | NM_001302946.2 | 8 | 1400 | NP_001289875.2 |
| NM_001367321.1 | NP_001354250.1 | |||
| NM_001367322.1 | 8 | 1759 | NP_001354251.1 | |
| NM_001367323.1 | 8 | 2488 | NP_001354252.1 | |
| NM_182916.3 | 8 | 1460 | NP_886552.3 | |
| NR_159934.1 | ||||
| NR_159935.1 | ||||
| NR_159936.1 | ||||
| NR_159937.1 | ||||
| NR_159938.1 | 7 | 1266 | ||
| NR_159939.1 | 7 | 1321 | ||
| NR_159940.1 | 7 | 1335 | ||
| NR_159941.1 | 8 | 2502 | ||
| XM_047448240.1 | XP_047304196.1 | |||
| XM_047448241.1 | XP_047304197.1 | |||
| AK290411.1 | BAF83100.1 | |||
| AL834397.1 | 8 | 1388 | CAD39059.1 | |
| BQ722111.1 | 1 | 311 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589371 | Chr.3:2170634 - 3594980 on Build GRCh38 | Loss |
|
| esv2422274 | Chr.3:2100396 - 3314907 on Build GRCh38 | Duplication |
|
| esv2751994 | Chr.3:2200757 - 3178886 on Build GRCh38 | Gain |
|
| nsv6375342 | Chr.3:2825261 - 3257855 on Build GRCh38 | Duplication |
|
| nsv521796 | Chr.3:3038825 - 3151219 on Build GRCh38 | Loss |
|
| dgv4636n100 | Chr.3:2825734 - 3271845 on Build GRCh38 | Gain |
|
| nsv4082884 | Chr.3:3089475 - 3252565 on Build GRCh38 | Deletion |
|
| nsv834604 | Chr.3:3147083 - 3361983 on Build GRCh38 | Gain |
|
Set Membership: |
|