Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02774808_cn
          See other CUBN CNV Assays ›
          Gene Symbol
          CUBN
          Assay Reference Genome
          Location

          Chr.10:16933176 on build GRCh38
          Cytoband
          Assay ID Hs02774808_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          8029

          Gene Name:

          cubilin

          Gene Aliases:

          IFCR, IGS, IGS1, MGA1, gp280

          Location:

          Chr.10:16823966-17129811 on Build GRCh38

          Assay Gene Location:

          Within Exon 43
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CUBN NM_001081.4 40 6081 NP_001072.2
          XM_011519708.3 40 6081 XP_011518010.1
          XM_011519709.3 14 2107 XP_011518011.1
          XM_011519710.3 14 2086 XP_011518012.1
          XM_011519711.4 13 2065 XP_011518013.1
          AB209880.1 13 2154 BAD93117.1
          AF034611.1 40 6061 AAC82612.1
          AK075228.1 BAG52089.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv470924 Chr.10:16864509 - 16951578 on Build GRCh38 Gain CUBN
          esv3891765 Chr.10:16809409 - 16977014 on Build GRCh38 Gain RSU1 CUBN
          nsv466758 Chr.10:16672467 - 16954685 on Build GRCh38 Gain RSU1 CUBN
          nsv831800 Chr.10:16765669 - 16949419 on Build GRCh38 Gain RSU1 CUBN
          nsv5481509 Chr.10:16808100 - 16978218 on Build GRCh38 Duplication RSU1 CUBN
          nsv5923254 Chr.10:16671260 - 16954786 on Build GRCh38 Duplication RSU1 CUBN
          esv3622477 Chr.10:16808207 - 16934688 on Build GRCh38 Gain RSU1 CUBN
          nsv5909086 Chr.10:16761962 - 17211984 on Build GRCh38 Deletion RSU1 TRDMT1 CUBN

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          tissue homeostasis
          lipid metabolic process
          endocytosis
          receptor-mediated endocytosis
          steroid metabolic process
          cholesterol metabolic process
          cobalamin metabolic process
          response to bacterium
          protein transport
          cobalamin transport
          lipoprotein transport
          establishment of localization in cell
          calcium ion binding
          protein binding
          cobalamin binding
          signaling receptor activity
          cargo receptor activity
          protein homodimerization activity
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline