Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02923187_cn
          See other RICTOR CNV Assays ›
          Gene Symbol
          RICTOR
          Assay Reference Genome
          Location

          Chr.5:38975577 on build GRCh38
          Cytoband
          Assay ID Hs02923187_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          253260

          Gene Name:

          RPTOR independent companion of MTOR complex 2

          Gene Aliases:

          AVO3, PIA, hAVO3

          Location:

          Chr.5:38937920-39074399 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 11 - Exon 12
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          RICTOR NM_001285439.2 NP_001272368.1
          NM_001285440.2 NP_001272369.1
          NM_001438007.1
          NM_001438049.1
          NM_001438246.1
          NM_001438247.1
          NM_001438248.1
          NM_001438249.1
          NM_152756.5 NP_689969.2
          XM_006714463.4 XP_006714526.1
          XM_011514005.3 XP_011512307.1
          XM_011514006.4 XP_011512308.1
          XM_017009311.2 XP_016864800.1
          XM_017009312.2 XP_016864801.1
          XM_047417068.1 XP_047273024.1
          XM_047417069.1 XP_047273025.1
          XM_047417070.1 XP_047273026.1
          AK128358.1
          AY515854.1 AAS79796.1
          BC137163.1 AAI37164.1
          BC137164.1 AAI37165.1
          BC144509.1 AAI44510.1
          CR749280.1 CAH18135.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv5636n100 Chr.5:38936517 - 39017379 on Build GRCh38 Loss OSMR RICTOR
          nsv529023 Chr.5:38955694 - 39119621 on Build GRCh38 Loss FYB1 RICTOR
          esv3604775 Chr.5:38961328 - 39069494 on Build GRCh38 Gain RICTOR

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          cytoskeleton organization
          lipid biosynthetic process
          embryo development ending in birth or egg hatching
          regulation of gene expression
          actin cytoskeleton organization
          positive regulation of cell growth
          positive regulation of cell migration
          positive regulation of actin filament polymerization
          cellular response to nutrient levels
          TOR signaling
          positive regulation of TOR signaling
          regulation of actin cytoskeleton organization
          TORC2 signaling
          negative regulation of apoptotic process
          protein kinase B signaling
          regulation of inflammatory response
          regulation of protein kinase B signaling
          positive regulation of protein kinase B signaling
          cGAS/STING signaling pathway
          negative regulation of cGAS/STING signaling pathway
          regulation of establishment of cell polarity
          nucleotide binding
          protein binding
          ATP binding
          zinc ion binding
          protein kinase binding
          ribosome binding
          protein serine/threonine kinase activator activity
          binding, bridging
          enzyme-substrate adaptor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline