Search
Search

Entrez Gene ID: | 5889 |
Gene Name: | RAD51 paralog C |
Gene Aliases: |
BROVCA3, FANCO, R51H3, RAD51L2 |
Location: |
Chr.17:58692538-58734342 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51C | NM_002876.3 | 2 | 368 | NP_002867.1 |
| NM_058216.2 | 2 | 368 | NP_478123.1 | |
| NR_103872.1 | 2 | 368 | ||
| NR_103873.1 | 2 | 265 | ||
| XM_006722001.3 | 2 | 339 | XP_006722064.1 | |
| XM_006722002.3 | 2 | 339 | XP_006722065.1 | |
| XM_006722004.3 | 2 | 261 | XP_006722067.1 | |
| XM_006722005.3 | 2 | 247 | XP_006722068.1 | |
| XM_011525092.2 | 2 | 467 | XP_011523394.1 | |
| XM_011525093.2 | 2 | 628 | XP_011523395.1 | |
| XM_011525094.2 | 2 | 316 | XP_011523396.1 | |
| XM_017024914.1 | 2 | 261 | XP_016880403.1 | |
| XM_017024915.1 | 2 | 613 | XP_016880404.1 | |
| XM_017024916.1 | 3 | 521 | XP_016880405.1 | |
| XM_017024917.1 | 2 | 246 | XP_016880406.1 | |
| XM_017024918.1 | 2 | 315 | XP_016880407.1 | |
| XM_017024919.1 | 2 | 628 | XP_016880408.1 | |
| AA304618.1 | 2 | 199 | ||
| AF029669.1 | 2 | 339 | AAC39604.1 | |
| AF029670.1 | 2 | 313 | AAC39605.1 | |
| AI424321.1 | 1 | 269 | ||
| AK290487.1 | 2 | 339 | ||
| AK303363.1 | 2 | 278 | ||
| BC093930.1 | 2 | 334 | AAH93930.1 | |
| BC101485.1 | 2 | 334 | AAI01486.1 | |
| BC107753.1 | 2 | 372 | AAI07754.1 | |
| BM987503.1 | 1 | 272 | ||
| BT007339.1 | 2 | 297 | AAP36003.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833498 | Chr.17:58652063 - 58815031 on Build GRCh38 | Loss |
|
| nsv1066945 | Chr.17:58650965 - 58704810 on Build GRCh38 | Loss |
|
| nsv428347 | Chr.17:58565015 - 58717588 on Build GRCh38 | Gain |
|
Set Membership: |
|