Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02969647_cn
          See other SERPINB2 CNV Assays ›
          Gene Symbol
          SERPINB2
          Assay Reference Genome
          Location

          Chr.18:63897222 on build GRCh38
          Cytoband
          Assay ID Hs02969647_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5055

          Gene Name:

          serpin family B member 2

          Gene Aliases:

          HsT1201, PAI, PAI-2, PAI2, PLANH2

          Location:

          Chr.18:63887705-63903888 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 4 - Intron 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SERPINB2 NM_001143818.2 NP_001137290.1
          NM_002575.3 NP_002566.1
          XM_024451192.2 XP_024306960.1
          A21238.1 CAA01535.1
          A32415.1 CAA02099.1
          AK313159.1 BAG35977.1
          BC012609.1 AAH12609.1
          J02685.1 AAA36413.1
          J03603.1 AAA60004.1
          M18082.1 AAA60006.1
          Y00630.1 CAA68666.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1066552 Chr.18:63147299 - 64608176 on Build GRCh38 Gain KDSR-DT SERPINB4 SERPINB2 LINC00305 SERPINB3 BCL2 SERPINB13 HMSD SERPINB12 VPS4B SERPINB8 KDSR LINC01924 SERPINB10 SERPINB11 SERPINB5 LINC01538 SERPINB7
          nsv6526003 Chr.18:63713430 - 64357552 on Build GRCh38 Deletion SERPINB8 SERPINB2 LINC00305 LINC01924 SERPINB10 SERPINB11 LINC01538 SERPINB7 HMSD
          nsv5514480 Chr.18:63714707 - 64278418 on Build GRCh38 Duplication SERPINB8 SERPINB2 LINC00305 LINC01924 SERPINB10 SERPINB11 LINC01538 SERPINB7 HMSD
          nsv833673 Chr.18:63897072 - 64097021 on Build GRCh38 Gain SERPINB8 SERPINB2 LINC00305 SERPINB10 HMSD
          nsv4532319 Chr.18:63817764 - 66099664 on Build GRCh38 Deletion SERPINB8 SERPINB2 LINC00305 LINC01924 SERPINB10 CDH7 LINC01538 HMSD
          nsv6519660 Chr.18:63146370 - 64608618 on Build GRCh38 Duplication KDSR-DT SERPINB4 SERPINB2 LINC00305 SERPINB3 BCL2 SERPINB13 HMSD SERPINB12 VPS4B SERPINB8 KDSR LINC01924 SERPINB10 SERPINB11 SERPINB5 LINC01538 SERPINB7
          esv21739 Chr.18:63715122 - 64278129 on Build GRCh38 Gain SERPINB8 SERPINB2 LINC00305 LINC01924 SERPINB10 SERPINB11 LINC01538 SERPINB7 HMSD
          esv3642785 Chr.18:63831255 - 63986077 on Build GRCh38 Gain SERPINB8 SERPINB2 SERPINB10 HMSD
          nsv1067311 Chr.18:63818011 - 66637685 on Build GRCh38 Loss SERPINB8 SERPINB2 LINC00305 CDH19 LINC01924 SERPINB10 CDH7 LINC01538 HMSD
          nsv6520851 Chr.18:63775316 - 64080261 on Build GRCh38 Duplication SERPINB8 SERPINB2 LINC00305 SERPINB10 SERPINB7 HMSD
          nsv525889 Chr.18:63845132 - 63948110 on Build GRCh38 Loss SERPINB2 SERPINB10
          esv2758730 Chr.18:63881807 - 63923851 on Build GRCh38 Loss SERPINB2 SERPINB10

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          fibrinolysis
          negative regulation of apoptotic process
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline