Search
Search

Entrez Gene ID: | 202309 |
Gene Name: | GRB2 binding adaptor protein, transmembrane |
Gene Aliases: |
C5orf29 |
Location: |
Chr.5:58491435-58497090 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAPT | NM_001304428.2 | 3 | 1656 | NP_001291357.1 |
| NM_001304429.2 | 3 | 1649 | NP_001291358.1 | |
| NM_001304431.2 | 3 | 1557 | NP_001291360.1 | |
| NM_152687.4 | 3 | 1494 | NP_689900.1 | |
| AK090960.1 | 3 | 1428 | BAC03557.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4844 | Chr.5:58476094 - 58509159 on Build GRCh38 | Insertion |
|
| nsv4523070 | Chr.5:57950170 - 58961173 on Build GRCh38 | Duplication |
|
| nsv5465476 | Chr.5:57931929 - 58692366 on Build GRCh38 | Duplication |
|
Set Membership: |
|