Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03039289_cn
          See other CAMK2D CNV Assays ›
          Gene Symbol
          CAMK2D
          Assay Reference Genome
          Location

          Chr.4:113453637 on build GRCh38
          Cytoband
          Assay ID Hs03039289_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          817

          Gene Name:

          calcium/calmodulin dependent protein kinase II delta

          Gene Aliases:

          CAMKD

          Location:

          Chr.4:113451032-113761738 on Build GRCh38

          Assay Gene Location:

          Within Exon 26
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CAMK2D NM_001221.4 19 3078 NP_001212.2
          NM_001321566.2 20 3120 NP_001308495.1
          NM_001321567.2 20 3128 NP_001308496.1
          NM_001321568.2 20 3057 NP_001308497.1
          NM_001321569.2 21 3180 NP_001308498.1
          NM_001321570.2 20 3084 NP_001308499.1
          NM_001321571.2 21 3180 NP_001308500.1
          NM_001321572.2 20 3084 NP_001308501.1
          NM_001321573.2 21 3153 NP_001308502.1
          NM_001321574.2 19 3075 NP_001308503.1
          NM_001321575.2 19 3024 NP_001308504.1
          NM_001321576.2 18 2979 NP_001308505.1
          NM_001321577.2 19 3021 NP_001308506.1
          NM_001321578.2 18 2951 NP_001308507.1
          NM_001321579.2 20 3128 NP_001308508.1
          NM_001321580.2 20 3138 NP_001308509.1
          NM_001321581.2 20 3125 NP_001308510.1
          NM_001321582.2 19 3015 NP_001308511.1
          NM_001321583.2 19 3068 NP_001308512.1
          NM_001321584.2 19 3021 NP_001308513.1
          NM_001321585.2 20 3118 NP_001308514.1
          NM_001321586.2 20 3081 NP_001308515.1
          NM_001321587.2 20 3057 NP_001308516.1
          NM_001321588.2 20 3117 NP_001308517.1
          NM_001321589.2 19 3068 NP_001308518.1
          NM_001321590.2 19 3024 NP_001308519.1
          NM_001321591.2 20 3101 NP_001308520.1
          NM_001396964.1 20 3120 NP_001383893.1
          NM_001396965.1 20 3111 NP_001383894.1
          NM_001396966.1 20 3108 NP_001383895.1
          NM_001396967.1 18 3023 NP_001383896.1
          NM_001399855.1 20 3125 NP_001386784.1
          NM_001399856.1 20 3125 NP_001386785.1
          NM_001399857.1 19 3012 NP_001386786.1
          NM_001399858.1 18 2982 NP_001386787.1
          NM_001399859.1 20 3179 NP_001386788.1
          NM_001399860.1 19 3083 NP_001386789.1
          NM_001399861.1 19 3083 NP_001386790.1
          NM_001399862.1 19 3124 NP_001386791.1
          NM_001399863.1 21 3157 NP_001386792.1
          NM_001399864.1 19 3052 NP_001386793.1
          NM_001399865.1 19 3076 NP_001386794.1
          NM_172127.3 18 3026 NP_742125.1
          NM_172128.3 18 2982 NP_742126.1
          XM_017008674.3 20 3081 XP_016864163.1
          AK025368.1 7 1466
          BX538071.1 1 2620
          BX647363.1 1 2777

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv830038 Chr.4:113289857 - 113470695 on Build GRCh38 Gain ANK2 CAMK2D
          nsv5547030 Chr.4:68996170 - 113894177 on Build GRCh38 Insertion AREG CDKL2 ADH1C PPP3CA UMLILO THAP6 LINC02173 ODAPH MANBA STPG2 H2AZ1 NEUROG2-AS1 INKILN HPSE NFKB1 GRSF1 PPA2 ADH4 MIR4452 WDFY3-AS1 GPRIN3 EMCN EIF4E MAPK10-AS1 LOC124900866 CXXC4 LINC02483 SNCA PF4 LOC102725220 HERC3 SCD5 EGF G3BP2 INTS12 JCHAIN SDAD1 HSD17B13 SPP1 THAP9 MIR575 SMARCAD1-DT SDAD1-AS1 NUDT9 DSPP DKK2 SMR3B UGT2B7 SLC4A4 ENPEP SNORD144 AP1AR FRAS1 OPRPN BMPR1B DMP1 PANCR RPS6P6 COQ2 LRIT3 EREG PCAT4 USO1 MIR302CHG ANTXR2 SPARCL1 MIR302A CENPE FAM13A GK2 MIR302D ODAM CABS1 PPBPP2 LOC105377329 NAP1L5 GSTCD METAP1 CSN1S2BP COL25A1 SULT1E1 BANK1 TIGD2 CXCL6 TRMT10A FAM47E-STBD1 CFI CXCL2 ELOVL6 CCNG2 CYP2U1-AS1 HERC6 SNORA101A H2AZ1-DT MRPL1 ADH1B CSN1S2AP KLHL8 LOC105377267 PPEF2 DDIT4L-AS1 CCDC158 GAR1-DT DMP1-AS1 LOC105377352 GAR1 HSP90AB3P SMR3A BMP2K-DT BDH2 DNAJB14 HSD17B11 ARHGEF38 ARHGEF38-IT1 BMP2K PPBP ALB LINC02428 UBE2D3-AS1 TSPAN5 TBCK NEUROG2 WDFY3-AS2 PLA2G12A SNORD161 LINC01218 MIR297 ADH5 HERC5 CXCL10 SOWAHB UGT2B11 FDCSP LOC105377276 BTC CAMK2D LINC02503 CCSER1 MIR548AH C4orf36 TIFA LOC102723704 UGT2B28 HPGDS CXCL8 IBSP PPM1K-DT UBE2D3 LOC124900855 SEPTIN11 MRPS18C UTP3 SLC9B2 CXCL1 MIR302B GPAT3 NKX6-1 TET2-AS1 SHROOM3 ANK2 SLC9B1 OSTC PRDM8 BMP3 MIR5705 CXCL13 THAP9-AS1 PIGY MIR576 C4orf54 AMTN SCARB2 LOC124900752 ZGRF1 AFF1-AS1 PAPSS1 LAMTOR3 MTHFD2L MIR367 MIR8082 CXCL9 SEC24B-AS1 PRDM8-AS1 TACR3 MIR1243 AIMP1 PITX2 LINC02945 LINC01094 HNRNPD-DT TET2 NFKB1-AS1 SEC31A PIGY-DT CYP2U1 ADAMTS3 NAA11 STBD1 ANKRD17-DT PRR27 LINC02994 UGT2B4 SGMS2 BMPR1B-DT HNRNPD TACR3-AS1 CXXC4-AS1 MIR3684 LOC101928893 STATH GRID2 MEPE RUFY3 LOC100421494 MIR8066 AFM LINC02562 HNRNPDL DCK ALPK1 ARHGAP24 ANKRD17 RASSF6 HELQ MCUB GSTCD-AS1 MOB1B MIR4450 PARM1-AS1 PYURF RRH RPL34-DT UGT2A2 SHROOM3-AS1 COL25A1-DT SMARCAD1 ABCG2 UNC5C-AS1 SEC24B LEF1 PKD2 PF4V1 RASGEF1B MIR1255A CSN2 LINC01088 LINC00575 CXCL5 LOC100507053 PRKG2-AS1 SULT1B1 TSPAN5-DT CSN1S1 MIR302C CXCL11 LINC01217 MUC7 LOC101929577 LOC124900868 NUP54 CASP6 ADH6 DAPP1 RPL34 LINC02267 FAM241A AFP ENOPH1 CFAP299 ABRAXAS1 NPNT CSN3 FAM13A-AS1 SLC10A6 LINC01216 PAQR3 COX18 RPS26P25 C4orf17 TMEM150C PARM1 SLC39A8 AMBN COPS4 PLAC8 ADH7 MAPK10 AP1AR-DT CHCHD2P7 ANXA3 ADH1A LNCPRESS2 PCNAP1 LARP7 SNORD143 LOC105377366 UGT2A1 ATOH1 LEF1-AS1 FGF5 WDFY3 HADH FAM47E STPG2-AS1 HTN3 PRKG2 EPGN PTPN13 ETNPPL GC NAAA CNOT6L PPP3CA-DT MMRN1 PDLIM5 LINC02499 MIR4451 PDHA2 ART3 GIMD1 CXCL3 CCNI TMSB4XP8 NPFFR2 AFF1 MTTP ENAM PPM1K LINC00989 UNC5C SNCA-AS1 RCHY1 LOC124900867 LIN54 CISD2 LOC101929621 SCPPPQ1 CDS1 HTN1 RAP1GDS1 DDIT4L

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          regulation of the force of heart contraction
          regulation of membrane depolarization
          regulation of transcription from RNA polymerase II promoter
          protein phosphorylation
          regulation of heart contraction
          positive regulation of cardiac muscle hypertrophy
          regulation of cell communication by electrical coupling
          positive regulation of cardiac muscle cell apoptotic process
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          endoplasmic reticulum calcium ion homeostasis
          regulation of neuronal synaptic plasticity
          relaxation of cardiac muscle
          long-term synaptic potentiation
          regulation of ryanodine-sensitive calcium-release channel activity
          regulation of cellular localization
          cellular response to calcium ion
          cardiac muscle cell contraction
          regulation of heart rate by cardiac conduction
          regulation of cardiac muscle cell action potential
          regulation of cardiac muscle cell action potential involved in regulation of contraction
          regulation of cell communication by electrical coupling involved in cardiac conduction
          regulation of relaxation of cardiac muscle
          negative regulation of sodium ion transmembrane transport
          regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
          regulation of protein localization to plasma membrane
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          protein binding
          calmodulin binding
          ATP binding
          kinase activity
          transferase activity
          sodium channel inhibitor activity
          titin binding
          identical protein binding
          protein homodimerization activity
          ion channel binding
          protein serine kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline