Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03086596_cn
          See other SUMF1 CNV Assays ›
          Gene Symbol
          SUMF1
          Assay Reference Genome
          Location

          Chr.3:4226834 on build GRCh38
          Cytoband
          Assay ID Hs03086596_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          285362

          Gene Name:

          sulfatase modifying factor 1

          Gene Aliases:

          AAPA3037, FGE, UNQ3037

          Location:

          Chr.3:4034486-4467269 on Build GRCh38

          Assay Gene Location:

          Within Intron 12
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SUMF1 XM_011533624.4 XP_011531926.1
          XM_017006252.3 XP_016861741.1
          XM_017006253.2 XP_016861742.1
          XM_017006254.3 XP_016861743.1
          AY358092.1 AAQ88459.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv1354e212 Chr.3:4125001 - 4271801 on Build GRCh38 Loss SUMF1
          esv2677838 Chr.3:4198342 - 4240523 on Build GRCh38 Deletion SUMF1
          nsv4077045 Chr.3:3819660 - 4285363 on Build GRCh38 Duplication LRRN1 SUMF1
          dgv4775n223 Chr.3:4122913 - 4241717 on Build GRCh38 Deletion SUMF1
          dgv4652n100 Chr.3:4104499 - 4295321 on Build GRCh38 Loss SUMF1
          dgv4773n223 Chr.3:4100085 - 4295563 on Build GRCh38 Deletion SUMF1
          nsv4083341 Chr.3:4069502 - 4294580 on Build GRCh38 Duplication SUMF1
          esv3893640 Chr.3:4196512 - 4421324 on Build GRCh38 Loss SETMAR SUMF1
          esv3568590 Chr.3:4199056 - 4268613 on Build GRCh38 Loss SUMF1
          dgv1355e212 Chr.3:4192873 - 4235682 on Build GRCh38 Loss SUMF1
          dgv339n206 Chr.3:4110265 - 4271913 on Build GRCh38 Deletion SUMF1
          nsv1160918 Chr.3:4198163 - 4244197 on Build GRCh38 Deletion SUMF1
          nsv6360911 Chr.3:4222496 - 4230608 on Build GRCh38 Deletion SUMF1
          nsv4075055 Chr.3:4171676 - 4239316 on Build GRCh38 Deletion SUMF1
          dgv1599n166 Chr.3:4113837 - 4271914 on Build GRCh38 Deletion SUMF1
          nsv6369325 Chr.3:4195374 - 4246193 on Build GRCh38 Deletion SUMF1
          esv3595089 Chr.3:4130643 - 4271936 on Build GRCh38 Loss SUMF1
          nsv5437255 Chr.3:4210000 - 4293000 on Build GRCh38 Deletion SUMF1
          dgv1596n166 Chr.3:4052064 - 4273252 on Build GRCh38 Deletion SUMF1
          esv3893631 Chr.3:4119651 - 4283616 on Build GRCh38 Loss SUMF1
          nsv4075421 Chr.3:4202373 - 4270755 on Build GRCh38 Deletion SUMF1
          dgv1603n166 Chr.3:4223615 - 4308772 on Build GRCh38 Deletion SETMAR SUMF1
          dgv8184n54 Chr.3:4140066 - 4242079 on Build GRCh38 Loss SUMF1
          nsv5993378 Chr.3:4198457 - 4240465 on Build GRCh38 Deletion SUMF1
          nsv6375114 Chr.3:4224484 - 4265171 on Build GRCh38 Deletion SUMF1
          dgv272n21 Chr.3:4080585 - 4319799 on Build GRCh38 Loss SETMAR SUMF1
          esv2763711 Chr.3:3921859 - 4227347 on Build GRCh38 Loss SUMF1
          nsv1008183 Chr.3:4221306 - 4237761 on Build GRCh38 Loss SUMF1
          dgv8174n54 Chr.3:4019253 - 4242079 on Build GRCh38 Loss SUMF1
          esv2759126 Chr.3:4096685 - 4378673 on Build GRCh38 Loss SETMAR SUMF1
          dgv744n145 Chr.3:4199999 - 4236672 on Build GRCh38 Deletion SUMF1
          nsv589450 Chr.3:4211535 - 4242079 on Build GRCh38 Loss SUMF1
          dgv4654n100 Chr.3:4123660 - 4237761 on Build GRCh38 Loss SUMF1
          nsv999405 Chr.3:4224848 - 4259411 on Build GRCh38 Loss SUMF1
          dgv1601n166 Chr.3:4172503 - 4285947 on Build GRCh38 Deletion SUMF1
          nsv589428 Chr.3:4084575 - 4283616 on Build GRCh38 Loss SUMF1
          dgv8171n54 Chr.3:3858107 - 4242079 on Build GRCh38 Loss SUMF1
          esv2724824 Chr.3:4198426 - 4240466 on Build GRCh38 Deletion SUMF1
          nsv4087272 Chr.3:4210624 - 4231254 on Build GRCh38 Deletion SUMF1
          nsv4083558 Chr.3:4023866 - 4313333 on Build GRCh38 Deletion SETMAR SUMF1
          dgv8185n54 Chr.3:4183047 - 4304082 on Build GRCh38 Loss SETMAR SUMF1
          nsv3681 Chr.3:4195647 - 4240502 on Build GRCh38 Deletion SUMF1
          nsv1007191 Chr.3:4195352 - 4237761 on Build GRCh38 Loss SUMF1
          nsv6372149 Chr.3:4206187 - 4253714 on Build GRCh38 Duplication SUMF1
          dgv592n27 Chr.3:4120923 - 4271051 on Build GRCh38 Loss SUMF1
          esv3595097 Chr.3:4195279 - 4246093 on Build GRCh38 Loss SUMF1
          esv3893630 Chr.3:4204149 - 4234996 on Build GRCh38 Loss SUMF1
          dgv340n206 Chr.3:4195376 - 4246193 on Build GRCh38 Deletion SUMF1
          nsv6360327 Chr.3:4204901 - 4228600 on Build GRCh38 Deletion SUMF1
          dgv8183n54 Chr.3:4106646 - 4271051 on Build GRCh38 Loss SUMF1
          nsv4085541 Chr.3:4170096 - 4535684 on Build GRCh38 Duplication SETMAR ITPR1 SUMF1 ITPR1-DT
          dgv1590n166 Chr.3:3929380 - 4240992 on Build GRCh38 Deletion SUMF1
          nsv6366515 Chr.3:4209110 - 4340569 on Build GRCh38 Deletion SETMAR SUMF1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          protein oxidation
          post-translational protein modification
          glycosphingolipid catabolic process
          protein binding
          oxidoreductase activity
          identical protein binding
          metal ion binding
          Formylglycine-generating oxidase activity
          cupric ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline