Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03105052_cn
          See other SUMF1 CNV Assays ›
          Gene Symbol
          SUMF1
          Assay Reference Genome
          Location

          Chr.3:4104732 on build GRCh38
          Cytoband
          Assay ID Hs03105052_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          285362

          Gene Name:

          sulfatase modifying factor 1

          Gene Aliases:

          AAPA3037, FGE, UNQ3037

          Location:

          Chr.3:4034486-4467269 on Build GRCh38

          Assay Gene Location:

          Within Intron 12
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SUMF1 XM_011533624.4 XP_011531926.1
          XM_017006252.3 XP_016861741.1
          XM_017006253.2 XP_016861742.1
          XM_017006254.3 XP_016861743.1
          AY358092.1 AAQ88459.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4077045 Chr.3:3819660 - 4285363 on Build GRCh38 Duplication LRRN1 SUMF1
          nsv6372587 Chr.3:4047010 - 4109380 on Build GRCh38 Deletion SUMF1
          nsv519512 Chr.3:4036970 - 4183047 on Build GRCh38 Loss SUMF1
          dgv4768n223 Chr.3:4066144 - 4176829 on Build GRCh38 Deletion SUMF1
          dgv4646n100 Chr.3:4046103 - 4107492 on Build GRCh38 Loss SUMF1
          esv3595086 Chr.3:4100349 - 4167284 on Build GRCh38 Loss SUMF1
          dgv4652n100 Chr.3:4104499 - 4295321 on Build GRCh38 Loss SUMF1
          dgv4766n223 Chr.3:4009545 - 4114078 on Build GRCh38 Deletion SUMF1
          dgv4773n223 Chr.3:4100085 - 4295563 on Build GRCh38 Deletion SUMF1
          nsv4083341 Chr.3:4069502 - 4294580 on Build GRCh38 Duplication SUMF1
          dgv1439n209 Chr.3:4030576 - 4133717 on Build GRCh38 Deletion SUMF1
          dgv4769n223 Chr.3:4083820 - 4143333 on Build GRCh38 Deletion SUMF1
          nsv528048 Chr.3:3957986 - 4171349 on Build GRCh38 Loss SUMF1
          dgv1595n166 Chr.3:4049381 - 4208377 on Build GRCh38 Deletion SUMF1
          dgv4641n100 Chr.3:3934721 - 4221511 on Build GRCh38 Loss SUMF1
          dgv1591n166 Chr.3:3962495 - 4192313 on Build GRCh38 Deletion SUMF1
          nsv4080499 Chr.3:4036329 - 4195193 on Build GRCh38 Deletion SUMF1
          dgv4648n100 Chr.3:4066953 - 4172280 on Build GRCh38 Loss SUMF1
          nsv589407 Chr.3:4000518 - 4107492 on Build GRCh38 Loss SUMF1
          nsv3115388 Chr.3:4079460 - 4128601 on Build GRCh38 Deletion SUMF1
          dgv4642n100 Chr.3:4019632 - 4112924 on Build GRCh38 Loss SUMF1
          nsv6371042 Chr.3:4103533 - 4156885 on Build GRCh38 Duplication SUMF1
          esv3893635 Chr.3:3899706 - 4211535 on Build GRCh38 Loss SUMF1
          dgv1592n166 Chr.3:3981800 - 4122959 on Build GRCh38 Deletion SUMF1
          dgv337n206 Chr.3:4076788 - 4218922 on Build GRCh38 Deletion SUMF1
          nsv460346 Chr.3:4093768 - 4198269 on Build GRCh38 Loss SUMF1
          dgv4765n223 Chr.3:4006276 - 4217513 on Build GRCh38 Deletion SUMF1
          nsv5434185 Chr.3:4100429 - 4167255 on Build GRCh38 Deletion SUMF1
          esv3595075 Chr.3:3957444 - 4160335 on Build GRCh38 Loss SUMF1
          dgv1593n166 Chr.3:4028884 - 4133718 on Build GRCh38 Deletion SUMF1
          nsv526916 Chr.3:4057135 - 4143342 on Build GRCh38 Loss SUMF1
          dgv4647n100 Chr.3:4054960 - 4141728 on Build GRCh38 Loss SUMF1
          dgv8173n54 Chr.3:3924671 - 4188112 on Build GRCh38 Loss SUMF1
          dgv1596n166 Chr.3:4052064 - 4273252 on Build GRCh38 Deletion SUMF1
          dgv4767n223 Chr.3:4064909 - 4107601 on Build GRCh38 Deletion SUMF1
          dgv8180n54 Chr.3:4087093 - 4124049 on Build GRCh38 Loss SUMF1
          nsv6373968 Chr.3:4070980 - 4205768 on Build GRCh38 Deletion SUMF1
          dgv8178n54 Chr.3:4051238 - 4107492 on Build GRCh38 Loss SUMF1
          nsv6364096 Chr.3:4054633 - 4136813 on Build GRCh38 Deletion SUMF1
          dgv865e214 Chr.3:4092194 - 4142900 on Build GRCh38 Loss SUMF1
          dgv4644n100 Chr.3:4038014 - 4210583 on Build GRCh38 Loss SUMF1
          dgv864e214 Chr.3:4084346 - 4218932 on Build GRCh38 Loss SUMF1
          esv2752006 Chr.3:4047125 - 4198166 on Build GRCh38 Loss SUMF1
          nsv589410 Chr.3:4021961 - 4163939 on Build GRCh38 Loss SUMF1
          nsv5449365 Chr.3:4073830 - 4120314 on Build GRCh38 Deletion SUMF1
          nsv5993373 Chr.3:4084855 - 4203810 on Build GRCh38 Deletion SUMF1
          dgv1353e212 Chr.3:4073038 - 4189176 on Build GRCh38 Loss SUMF1
          dgv4651n100 Chr.3:4099559 - 4172280 on Build GRCh38 Loss SUMF1
          dgv8179n54 Chr.3:4079225 - 4205501 on Build GRCh38 Loss SUMF1
          esv3893628 Chr.3:4080585 - 4204149 on Build GRCh38 Loss SUMF1
          dgv4762n223 Chr.3:3857761 - 4141396 on Build GRCh38 Deletion SUMF1
          esv2763711 Chr.3:3921859 - 4227347 on Build GRCh38 Loss SUMF1
          dgv588n27 Chr.3:4019253 - 4218633 on Build GRCh38 Loss SUMF1
          esv2759126 Chr.3:4096685 - 4378673 on Build GRCh38 Loss SETMAR SUMF1
          nsv5898647 Chr.3:4103409 - 4121415 on Build GRCh38 Deletion SUMF1
          dgv1438n209 Chr.3:4002099 - 4219002 on Build GRCh38 Deletion SUMF1
          dgv1594n166 Chr.3:4040749 - 4109380 on Build GRCh38 Deletion SUMF1
          dgv8181n54 Chr.3:4102189 - 4140066 on Build GRCh38 Loss SUMF1
          dgv1598n166 Chr.3:4099316 - 4188316 on Build GRCh38 Deletion SUMF1
          nsv460337 Chr.3:4032349 - 4124049 on Build GRCh38 Loss SUMF1
          dgv4772n223 Chr.3:4098929 - 4113074 on Build GRCh38 Deletion SUMF1
          nsv589428 Chr.3:4084575 - 4283616 on Build GRCh38 Loss SUMF1
          nsv1002707 Chr.3:3858107 - 4139829 on Build GRCh38 Loss SUMF1
          nsv3114062 Chr.3:4084850 - 4199999 on Build GRCh38 Deletion SUMF1
          dgv338n206 Chr.3:4092357 - 4142769 on Build GRCh38 Deletion SUMF1
          nsv5449395 Chr.3:4030578 - 4163790 on Build GRCh38 Deletion SUMF1
          nsv4083558 Chr.3:4023866 - 4313333 on Build GRCh38 Deletion SETMAR SUMF1
          nsv589435 Chr.3:4103854 - 4107492 on Build GRCh38 Loss SUMF1
          dgv1597n166 Chr.3:4061464 - 4120314 on Build GRCh38 Deletion SUMF1
          esv3595081 Chr.3:4074541 - 4121014 on Build GRCh38 Loss SUMF1
          nsv5890732 Chr.3:4100427 - 4167254 on Build GRCh38 Deletion SUMF1
          esv3568575 Chr.3:3935472 - 4195417 on Build GRCh38 Loss SUMF1
          esv2760789 Chr.3:4024902 - 4194031 on Build GRCh38 Loss SUMF1
          dgv4650n100 Chr.3:4079539 - 4189324 on Build GRCh38 Loss SUMF1
          dgv4764n223 Chr.3:3933489 - 4219482 on Build GRCh38 Deletion SUMF1
          nsv5435439 Chr.3:3957458 - 4160267 on Build GRCh38 Deletion SUMF1
          dgv4771n223 Chr.3:4098635 - 4167255 on Build GRCh38 Deletion SUMF1
          nsv3169507 Chr.3:3859793 - 4143342 on Build GRCh38 Deletion SUMF1
          esv2752004 Chr.3:3979832 - 4163939 on Build GRCh38 Loss SUMF1
          dgv1590n166 Chr.3:3929380 - 4240992 on Build GRCh38 Deletion SUMF1
          esv2676337 Chr.3:4099408 - 4142779 on Build GRCh38 Deletion SUMF1
          nsv1155948 Chr.3:4092560 - 4132155 on Build GRCh38 Deletion SUMF1

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          protein oxidation
          post-translational protein modification
          glycosphingolipid catabolic process
          protein binding
          oxidoreductase activity
          identical protein binding
          metal ion binding
          Formylglycine-generating oxidase activity
          cupric ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline