Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03179648_cn
          See other IFIT1 CNV Assays ›
          Gene Symbol
          IFIT1
          Assay Reference Genome
          Location

          Chr.10:89403634 on build GRCh38
          Cytoband
          Assay ID Hs03179648_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          3434

          Gene Name:

          interferon induced protein with tetratricopeptide repeats 1

          Gene Aliases:

          C56, G10P1, IFI-56, IFI-56K, IFI56, IFIT-1, IFNAI1, ISG56, P56, RNM561

          Location:

          Chr.10:89392623-89406487 on Build GRCh38

          Assay Gene Location:

          Within Exon 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          IFIT1 NM_001270927.2 3 1546 NP_001257856.1
          NM_001270928.2 3 1563 NP_001257857.1
          NM_001270929.2 3 1558 NP_001257858.1
          NM_001270930.2 3 1684 NP_001257859.1
          NM_001548.5 2 1449 NP_001539.3
          AK092813.1 2 2554 BAG52612.1
          AK308688.1 3 1527
          AK314588.1 2 1449 BAG37163.1
          BC007091.1 2 1451 AAH07091.1
          BT006667.1 1 1359 AAP35313.1
          CA306581.1 1 370
          X03557.1 2 1423 CAA27244.1

          Target Gene Details

          Entrez Gene ID:

          3988

          Gene Name:

          lipase A, lysosomal acid type

          Gene Aliases:

          CESD, LAL

          Location:

          Chr.10:89213572-89414679 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LIPA NM_001440821.1
          NM_001440822.1
          NM_001440823.1
          NM_001440824.1
          NM_001440831.1
          NM_001440839.1
          AK096406.1 BAG53281.1
          AK125193.1 BAC86078.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv34085 Chr.10:89306192 - 89414418 on Build GRCh38 Loss LIPA IFIT3 IFIT1B IFIT1 IFIT2
          nsv6454393 Chr.10:89381814 - 89442315 on Build GRCh38 Deletion LIPA SLC16A12 IFIT5 IFIT1B IFIT1

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Exonic Intronic Non-exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          response to virus
          intracellular transport of viral protein in host cell
          positive regulation of viral genome replication
          negative regulation of viral genome replication
          innate immune response
          defense response to virus
          response to other organism
          cellular response to type I interferon
          cellular response to exogenous dsRNA
          antiviral innate immune response
          negative regulation of viral translation
          mitotic cell cycle
          cell morphogenesis
          retinoid metabolic process
          liver development
          endothelial cell proliferation
          response to dietary excess
          hematopoietic progenitor cell differentiation
          acute inflammatory response
          respiratory burst involved in inflammatory response
          glucose metabolic process
          glycolytic process
          lipid metabolic process
          fatty acid metabolic process
          neutral lipid metabolic process
          triglyceride metabolic process
          cholesterol biosynthetic process
          ATP biosynthetic process
          vitamin A metabolic process
          endocytosis
          inflammatory response
          mitochondrion organization
          lysosome organization
          small GTPase mediated signal transduction
          cholesterol metabolic process
          cell proliferation
          endosome to lysosome transport
          determination of adult lifespan
          lipid biosynthetic process
          response to cold
          response to xenobiotic stimulus
          gene expression
          cholesterol storage
          programmed cell death
          lipid catabolic process
          sterol metabolic process
          lipid storage
          hemopoiesis
          myeloid cell differentiation
          T cell differentiation
          cholesterol transport
          lung development
          defecation
          TOR signaling
          myeloid cell apoptotic process
          response to vitamin A
          cholesterol efflux
          response to lipid
          low-density lipoprotein particle clearance
          common myeloid progenitor cell proliferation
          T cell proliferation
          lipoprotein catabolic process
          regulation of membrane potential
          spleen development
          bone marrow development
          tissue remodeling
          multicellular organismal homeostasis
          homeostasis of number of cells
          homeostasis of number of cells within a tissue
          epithelial cell proliferation
          positive regulation of T cell receptor signaling pathway
          regulation of mitochondrial membrane potential
          lipid homeostasis
          adipose tissue development
          blood vessel endothelial cell differentiation
          macrophage homeostasis
          T cell apoptotic process
          fat cell proliferation
          triglyceride-rich lipoprotein particle clearance
          cell proliferation in bone marrow
          liver morphogenesis
          energy homeostasis
          lipid import into cell
          multicellular organismal-level chemical homeostasis
          response to rapamycin
          reactive oxygen species biosynthetic process
          adaptive thermogenesis
          RNA binding
          enzyme inhibitor activity
          protein binding
          RNA sequestering activity
          sterol esterase activity
          lipase activity
          hydrolase activity
          hydrolase activity, acting on ester bonds
          carboxylic ester hydrolase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline