Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03183226_cn
          See other H2BC15 CNV Assays ›
          Gene Symbol
          H2BC15
          Assay Reference Genome
          Location

          Chr.6:27838669 on build GRCh38
          Cytoband
          Assay ID Hs03183226_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          8341

          Gene Name:

          H2B clustered histone 15

          Gene Aliases:

          H2B/d, H2BFD, HIST1H2BN

          Location:

          Chr.6:27838569-27839110 on Build GRCh38

          Assay Gene Location:

          Within Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          H2BC15 NM_003520.4 1 101 NP_003511.1
          AK312228.1 1 117 BAG35161.1
          BC101411.1 AAI01412.1
          BC101412.1 AAI01413.1
          BC101413.1 AAI01414.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3608441 Chr.6:27827497 - 27839980 on Build GRCh38 Gain H4C12 H2AC15 H2BC15
          nsv5462086 Chr.6:27798000 - 27892000 on Build GRCh38 Duplication H3C10 H1-5 H2AC16 H3C11 H4C12 H2BC15 H2AC14 H2AC13 H3C12 H4C11 H4C13 H2BC14 H2BC13 H2AC15
          nsv4120990 Chr.6:27828961 - 27841298 on Build GRCh38 Deletion H4C12 H2AC15 H2BC15
          nsv5466550 Chr.6:27711907 - 27849290 on Build GRCh38 Duplication H2AC13 LOC100131289 H3C10 H4C11 H2BC14 H4C12 H2BC13 H2AC15 H2BC15 H2AC14
          nsv6398741 Chr.6:27314958 - 28461291 on Build GRCh38 Duplication ZSCAN26 LINC01012 LOC100131289 ZSCAN9 H1-5 ZNF184 ZKSCAN8 H2AC16 H3C11 ZKSCAN8P1 H4C12 H2AC14 VN1R10P ZSCAN23 PGBD1 ZSCAN16-AS1 H3C12 H4C13 H2BC14 ZSCAN12 ZSCAN16 ZNF204P ZNF391 ZNF165 TOB2P1 H3C10 OR2B6 ZKSCAN4 H2BC15 H2AC17 ZSCAN31 OR2B8 H2AC13 H4C11 NKAPL H2BC13 H2AC15 ZSCAN12P1 OR2B2 ZKSCAN3 H2BC17
          nsv6407191 Chr.6:27684847 - 27902588 on Build GRCh38 Duplication LINC01012 LOC100131289 H3C10 H1-5 H2AC16 H3C11 H4C12 H2BC15 H2AC14 H2AC17 H2AC13 H3C12 H4C11 H4C13 H2BC14 H2BC13 H2AC15 H2BC17
          nsv6400159 Chr.6:27753532 - 28002885 on Build GRCh38 Duplication LOC100131289 H3C10 H1-5 H2AC16 H3C11 H4C12 OR2B6 H2BC15 H2AC14 H2AC17 H2AC13 H3C12 H4C11 H4C13 H2BC14 H2BC13 H2AC15 OR2B2 H2BC17
          nsv4434899 Chr.6:27807223 - 27839222 on Build GRCh38 Duplication H2AC13 H3C10 H4C11 H2BC14 H4C12 H2BC13 H2AC15 H2BC15 H2AC14
          nsv969367 Chr.6:27838096 - 27839734 on Build GRCh38 Duplication H2AC15 H2BC15

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          DNA binding
          structural constituent of chromatin
          protein heterodimerization activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline