Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03191961_cn
          See other PPP1R14B CNV Assays ›
          Gene Symbol
          PPP1R14B
          Assay Reference Genome
          Location

          Chr.11:64244465 on build GRCh38
          Cytoband
          Assay ID Hs03191961_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          26472

          Gene Name:

          protein phosphatase 1 regulatory inhibitor subunit 14B

          Gene Aliases:

          PHI-1, PLCB3N, PNG, SOM172

          Location:

          Chr.11:64244479-64246943 on Build GRCh38

          Assay Gene Location:

          Overlaps - Exon 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PPP1R14B NM_138689.3 NP_619634.1
          BC014522.2 AAH14522.3
          BC094817.1 AAH94817.1
          X91195.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1159904 Chr.11:64150129 - 64279169 on Build GRCh38 Deletion DNAJC4 MACROD1 FKBP2 VEGFB LOC114841035 PPP1R14B-AS1 PLCB3 TRPT1 NUDT22 STIP1 BAD FERMT3 GPR137 PPP1R14B
          nsv951010 Chr.11:63942829 - 64352128 on Build GRCh38 Deletion FLRT1 DNAJC4 CATSPERZ VEGFB LOC114841035 PPP1R14B-AS1 PLCB3 TRPT1 NUDT22 STIP1 PRDX5 CCDC88B GPR137 NAA40 MACROD1 FKBP2 COX8A ESRRA KCNK4-CATSPERZ KCNK4 TRMT112 BAD FERMT3 LOC102723878 PPP1R14B MIR7155 OTUB1
          dgv1963n54 Chr.11:64174262 - 64329761 on Build GRCh38 Loss DNAJC4 CATSPERZ FKBP2 VEGFB ESRRA LOC114841035 KCNK4-CATSPERZ PPP1R14B-AS1 PLCB3 TRPT1 NUDT22 KCNK4 TRMT112 STIP1 BAD FERMT3 PRDX5 LOC102723878 GPR137 PPP1R14B
          nsv555187 Chr.11:64185649 - 64260416 on Build GRCh38 Loss DNAJC4 PPP1R14B-AS1 PLCB3 TRPT1 NUDT22 FKBP2 STIP1 VEGFB FERMT3 LOC114841035 PPP1R14B
          nsv555184 Chr.11:64156974 - 64275525 on Build GRCh38 Loss DNAJC4 MACROD1 FKBP2 VEGFB LOC114841035 PPP1R14B-AS1 PLCB3 TRPT1 NUDT22 STIP1 BAD FERMT3 GPR137 PPP1R14B
          nsv832186 Chr.11:64204788 - 64402587 on Build GRCh38 Loss DNAJC4 CATSPERZ FKBP2 VEGFB ESRRA LOC114841035 KCNK4-CATSPERZ PPP1R14B-AS1 PLCB3 TRPT1 NUDT22 KCNK4 TRMT112 MIR1237 BAD FERMT3 PRDX5 LOC102723878 CCDC88B GPR137 RPS6KA4 PPP1R14B MIR7155
          esv24861 Chr.11:64233793 - 64250642 on Build GRCh38 Gain DNAJC4 PPP1R14B-AS1 FKBP2 VEGFB LOC114841035 PPP1R14B
          nsv983032 Chr.11:64243929 - 64247666 on Build GRCh38 Duplication PPP1R14B-AS1 FKBP2 LOC114841035 PPP1R14B
          nsv469961 Chr.11:64055468 - 64318495 on Build GRCh38 Loss FLRT1 DNAJC4 CATSPERZ MACROD1 FKBP2 VEGFB ESRRA LOC114841035 KCNK4-CATSPERZ PPP1R14B-AS1 PLCB3 TRPT1 NUDT22 KCNK4 TRMT112 STIP1 BAD FERMT3 PRDX5 GPR137 PPP1R14B

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Non-exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response
          protein phosphatase inhibitor activity
          protein serine/threonine phosphatase inhibitor activity
          protein binding
          phosphatase inhibitor activity
          protein phosphatase regulator activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline