Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03192321_cn
          See other PRSS1 CNV Assays ›
          Gene Symbol
          PRSS1
          Assay Reference Genome
          Location

          Chr.7:142753074 on build GRCh38
          Cytoband
          Assay ID Hs03192321_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5644

          Gene Name:

          serine protease 1

          Gene Aliases:

          TRP1, TRY1, TRY4, TRYP1

          Location:

          Chr.7:142749472-142753072 on Build GRCh38

          Assay Gene Location:

          Overlaps - Exon 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PRSS1 NM_002769.5 NP_002760.1
          NR_172947.1
          NR_172948.1
          NR_172949.1
          NR_172950.1
          NR_172951.1
          AA835201.1
          AB306150.1 BAF94843.1
          AB306249.1 BAF94942.1
          AK312199.1 BAG35132.1
          AY006147.1 AAG15650.1
          BC128226.1 AAI28227.1
          CA948204.1
          CR627200.1
          DQ778083.1 ABI63347.1
          M22612.1 AAA61231.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv11644n54 Chr.7:142314978 - 142796623 on Build GRCh38 Loss TRBV12-1 MIR11400 TRBJ2-2 TRBV7-1 TRBV7-2 TRBV23-1 TRBV27 TRBV16 TRBV12-2 TRBC1 TRBV21-1 PRSS2 PRSS1 TRBV6-7 TRBV5-4 TRBV19 TRBV6-5
          nsv818563 Chr.7:142686711 - 142784320 on Build GRCh38 Loss PRSS2 PRSS1 TRBV27
          esv34617 Chr.7:142641736 - 142783765 on Build GRCh38 Loss PRSS2 PRSS1 TRBV23-1 TRBV27
          nsv1029719 Chr.7:142535861 - 142763407 on Build GRCh38 Loss TRBV21-1 PRSS1 TRBV19 TRBV23-1 TRBV27 TRBV16
          nsv831167 Chr.7:142693447 - 142815764 on Build GRCh38 Gain+Loss TRBC1 PRSS2 TRBJ2-2 PRSS1 TRBV27
          dgv1928n209 Chr.7:142747768 - 142774543 on Build GRCh38 Duplication PRSS2 PRSS1
          nsv6142469 Chr.7:142721154 - 142795710 on Build GRCh38 Deletion TRBC1 PRSS2 PRSS1
          nsv3220567 Chr.7:142720119 - 142783415 on Build GRCh38 Insertion PRSS2 PRSS1
          esv2759569 Chr.7:142434958 - 143027060 on Build GRCh38 Loss MIR11400 TRBJ2-2 TRPV6 TRBV23-1 TRBV27 TRBV16 TRBV12-2 TRBC1 TRBV21-1 KEL PRSS2 PRSS1 EPHB6 TRBV6-7 TRBV5-4 TRBV19 TRPV5 TRBV6-5 LLCFC1 OR9A2
          dgv490n206 Chr.7:142313662 - 142797120 on Build GRCh38 Deletion TRBV12-1 MIR11400 TRBJ2-2 TRBV7-1 TRBV7-2 TRBV23-1 TRBV27 TRBV16 TRBV12-2 TRBC1 TRBV21-1 PRSS2 PRSS1 TRBV6-7 TRBV5-4 TRBV19 TRBV6-5
          dgv1968e212 Chr.7:142705603 - 142802013 on Build GRCh38 Gain TRBC1 PRSS2 TRBJ2-2 PRSS1 TRBV27
          nsv831166 Chr.7:142618692 - 142768033 on Build GRCh38 Gain+Loss TRBV21-1 PRSS1 TRBV19 TRBV23-1 TRBV27
          nsv1021910 Chr.7:142705687 - 142768033 on Build GRCh38 Gain PRSS1 TRBV27
          dgv7150n223 Chr.7:142746701 - 142818500 on Build GRCh38 Duplication TRBC1 PRSS2 TRBJ2-2 PRSS1
          nsv6014462 Chr.7:142657213 - 142796373 on Build GRCh38 Deletion TRBC1 PRSS2 PRSS1 TRBV27
          nsv1018286 Chr.7:142236996 - 142767091 on Build GRCh38 Gain TRBV12-1 MIR11400 TRBV3-1 TRBV7-1 TRBV7-2 TRBV23-1 TRBV27 TRBV16 TRBV12-2 PRSS58 MOXD2P TRBV21-1 PRSS1 TRBV6-7 TRBV5-4 PRSS59P TRBV19 TRBV6-5
          dgv491n206 Chr.7:142560927 - 142796849 on Build GRCh38 Deletion TRBC1 TRBV21-1 PRSS2 TRBJ2-2 PRSS1 TRBV19 TRBV23-1 TRBV27 TRBV16
          nsv1024031 Chr.7:142694837 - 142768033 on Build GRCh38 Loss PRSS1 TRBV27
          nsv5920229 Chr.7:142544684 - 142795707 on Build GRCh38 Deletion TRBC1 TRBV21-1 PRSS2 PRSS1 TRBV19 TRBV23-1 TRBV27 TRBV16
          dgv841n27 Chr.7:142625892 - 142809818 on Build GRCh38 Loss TRBC1 TRBV21-1 PRSS2 TRBJ2-2 PRSS1 TRBV23-1 TRBV27
          nsv819176 Chr.7:142715598 - 142761122 on Build GRCh38 Loss PRSS1 TRBV27
          nsv1111346 Chr.7:142751496 - 142768000 on Build GRCh38 Tandem Duplication PRSS1
          dgv1926n209 Chr.7:142637379 - 142797453 on Build GRCh38 Deletion TRBC1 TRBV21-1 PRSS2 TRBJ2-2 PRSS1 TRBV23-1 TRBV27
          dgv1969e212 Chr.7:142719410 - 142785829 on Build GRCh38 Loss PRSS2 PRSS1
          nsv824336 Chr.7:142637349 - 142796031 on Build GRCh38 Loss TRBC1 TRBV21-1 PRSS2 PRSS1 TRBV23-1 TRBV27
          esv2735271 Chr.7:142747357 - 142758232 on Build GRCh38 Deletion PRSS1
          nsv1028035 Chr.7:142706976 - 142795888 on Build GRCh38 Loss TRBC1 PRSS2 PRSS1 TRBV27
          nsv5917069 Chr.7:142715859 - 142797009 on Build GRCh38 Deletion TRBC1 PRSS2 TRBJ2-2 PRSS1 TRBV27
          dgv7152n223 Chr.7:142747501 - 142760200 on Build GRCh38 Duplication PRSS1
          nsv6420341 Chr.7:142392405 - 142797003 on Build GRCh38 Deletion TRBV12-1 MIR11400 TRBJ2-2 TRBV23-1 TRBV27 TRBV16 TRBV12-2 TRBC1 TRBV21-1 PRSS2 PRSS1 TRBV6-7 TRBV5-4 TRBV19 TRBV6-5
          nsv515039 Chr.7:142750239 - 142757887 on Build GRCh38 Gain PRSS1
          esv3615288 Chr.7:142717153 - 142754210 on Build GRCh38 Loss PRSS1
          esv3576450 Chr.7:142634755 - 142785829 on Build GRCh38 Gain TRBV21-1 PRSS2 PRSS1 TRBV23-1 TRBV27
          dgv1925n209 Chr.7:142313665 - 142797449 on Build GRCh38 Deletion TRBV12-1 MIR11400 TRBJ2-2 TRBV7-1 TRBV7-2 TRBV23-1 TRBV27 TRBV16 TRBV12-2 TRBC1 TRBV21-1 PRSS2 PRSS1 TRBV6-7 TRBV5-4 TRBV19 TRBV6-5
          nsv6141900 Chr.7:142748779 - 142762779 on Build GRCh38 Duplication PRSS1
          dgv7151n223 Chr.7:142746897 - 142774544 on Build GRCh38 Duplication PRSS2 PRSS1

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Non-exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          digestion
          extracellular matrix disassembly
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline