Search
Search

Entrez Gene ID: | 339175 |
Gene Name: | methyltransferase 2A, tRNA N3-cytidine |
Gene Aliases: |
METTL2 |
Location: |
Chr.17:62423897-62453385 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL2A | NM_181725.4 | NP_859076.3 | ||
| AF161384.1 | AAF28944.1 | |||
| AK000991.1 | BAG50845.1 | |||
| AK022822.1 | BAB14260.1 | |||
| BC006985.1 | AAH06985.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3640974 | Chr.17:62407702 - 62560090 on Build GRCh38 | Gain |
|
| nsv5938015 | Chr.17:62447169 - 62462823 on Build GRCh38 | Deletion |
|
| nsv962358 | Chr.17:62413853 - 62449107 on Build GRCh38 | Duplication |
|
| nsv6032263 | Chr.17:62445520 - 62451622 on Build GRCh38 | Duplication |
|
Set Membership: |
|