Search
Search

Entrez Gene ID: | 1587 |
Gene Name: | ADAM metallopeptidase domain 3A (pseudogene) |
Gene Aliases: |
ADAM3, CYRN1, tMDCI |
Location: |
Chr.8:39451045-39522989 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM3A | NR_001569.3 | |||
| NR_024106.2 | ||||
| NR_024107.2 | ||||
| NR_073423.1 | ||||
| AJ005372.1 | ||||
| AK302269.1 | BAG63615.1 | |||
| AK308250.1 | ||||
| BC025386.1 | ||||
| BC042979.2 | ||||
| X89654.1 | ||||
| X89655.1 | ||||
| X89656.1 | ||||
| X89657.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv21867 | Chr.8:39374143 - 39529900 on Build GRCh38 | Gain+Loss |
|
| esv2764074 | Chr.8:39368221 - 39546485 on Build GRCh38 | Gain+Loss |
|
| dgv12093n54 | Chr.8:39365305 - 39540503 on Build GRCh38 | Gain |
|
| nsv6163 | Chr.8:39368556 - 39533370 on Build GRCh38 | Deletion |
|
| dgv4e221 | Chr.8:39375220 - 39520881 on Build GRCh38 | Loss |
|
| dgv2066e212 | Chr.8:39356430 - 39531484 on Build GRCh38 | Gain |
|
| esv995573 | Chr.8:39373591 - 39530281 on Build GRCh38 | Deletion |
|
| dgv7174n100 | Chr.8:39378072 - 39531593 on Build GRCh38 | Loss |
|
| dgv12110n54 | Chr.8:39483411 - 39532700 on Build GRCh38 | Loss |
|
| dgv67e213 | Chr.8:39378084 - 39531605 on Build GRCh38 | Loss |
|
| dgv12107n54 | Chr.8:39451651 - 39499982 on Build GRCh38 | Loss |
|
| nsv1074232 | Chr.8:39374535 - 39529881 on Build GRCh38 | Deletion |
|
| dgv7167n100 | Chr.8:39368209 - 39505415 on Build GRCh38 | Gain |
|
| esv3891382 | Chr.8:39374115 - 39532700 on Build GRCh38 | Loss |
|
| dgv1174n145 | Chr.8:39411668 - 39528445 on Build GRCh38 | Deletion |
|
| nsv5569845 | Chr.8:39374555 - 39529708 on Build GRCh38 | Deletion |
|
| dgv1176n145 | Chr.8:39478017 - 39528445 on Build GRCh38 | Deletion |
|
| dgv12109n54 | Chr.8:39482960 - 39511604 on Build GRCh38 | Loss |
|
| nsv831297 | Chr.8:39443602 - 39577174 on Build GRCh38 | Gain |
|
| dgv12105n54 | Chr.8:39444480 - 39541538 on Build GRCh38 | Loss |
|
| nsv511391 | Chr.8:39386829 - 39541538 on Build GRCh38 | Gain |
|
| dgv7166n100 | Chr.8:39345917 - 39552463 on Build GRCh38 | Gain |
|
| nsv819496 | Chr.8:39371531 - 39529890 on Build GRCh38 | Gain |
|
| dgv285n97 | Chr.8:39368830 - 39546375 on Build GRCh38 | Deletion |
|
| nsv8331 | Chr.8:39374150 - 39531387 on Build GRCh38 | Gain+Loss |
|
| dgv181e180 | Chr.8:39374481 - 39528780 on Build GRCh38 | Loss |
|
| nsv951129 | Chr.8:39373982 - 39529581 on Build GRCh38 | Deletion |
|
| esv3572691 | Chr.8:39461177 - 39526818 on Build GRCh38 | Loss |
|
| dgv12102n54 | Chr.8:39380149 - 39499982 on Build GRCh38 | Gain+Loss |
|
| nsv611048 | Chr.8:39416742 - 39499982 on Build GRCh38 | Gain |
|
| nsv428198 | Chr.8:39211790 - 39548205 on Build GRCh38 | Gain |
|
| nsv1021473 | Chr.8:39464301 - 39527663 on Build GRCh38 | Gain |
|
| nsv3114848 | Chr.8:39487501 - 39494202 on Build GRCh38 | Deletion |
|
| nsv3116152 | Chr.8:39476702 - 39501228 on Build GRCh38 | Deletion |
|
| dgv7170n100 | Chr.8:39372652 - 39540213 on Build GRCh38 | Gain+Loss |
|
| esv2761164 | Chr.8:39372664 - 39531605 on Build GRCh38 | Loss |
|
| dgv7318n223 | Chr.8:39359801 - 39578400 on Build GRCh38 | Deletion |
|
| dgv1171n145 | Chr.8:39368796 - 39530975 on Build GRCh38 | Deletion |
|
| dgv146n17 | Chr.8:39367449 - 39550909 on Build GRCh38 | Loss |
|
| esv3616947 | Chr.8:39338306 - 39531711 on Build GRCh38 | Loss |
|
| esv2759608 | Chr.8:39316760 - 39740120 on Build GRCh38 | Gain+Loss |
|
| dgv12091n54 | Chr.8:39349476 - 39498641 on Build GRCh38 | Gain |
|
| nsv611072 | Chr.8:39483882 - 39529660 on Build GRCh38 | Gain+Loss |
|
| dgv1179n67 | Chr.8:39370204 - 39531195 on Build GRCh38 | Loss |
|
| esv3572692 | Chr.8:39464300 - 39498857 on Build GRCh38 | Loss |
|
| dgv7178n100 | Chr.8:39426673 - 39511377 on Build GRCh38 | Loss |
|
| dgv7181n100 | Chr.8:39426730 - 39531593 on Build GRCh38 | Loss |
|
| dgv9140n152 | Chr.8:39337831 - 39604557 on Build GRCh38 | Deletion |
|
| dgv7175n100 | Chr.8:39389144 - 39552463 on Build GRCh38 | Gain |
|
| nsv611050 | Chr.8:39432224 - 39496421 on Build GRCh38 | Loss |
|
| dgv7171n100 | Chr.8:39372794 - 39499344 on Build GRCh38 | Loss |
|
| nsv508503 | Chr.8:39368506 - 39521967 on Build GRCh38 | Deletion |
|
| nsv611064 | Chr.8:39472638 - 39520881 on Build GRCh38 | Loss |
|
| dgv1177n145 | Chr.8:39480230 - 39503164 on Build GRCh38 | Deletion |
|
| esv2736900 | Chr.8:39374432 - 39529697 on Build GRCh38 | Deletion |
|
| dgv7182n100 | Chr.8:39481989 - 39540213 on Build GRCh38 | Loss |
|
| dgv2068e212 | Chr.8:39389579 - 39531247 on Build GRCh38 | Loss |
|
| esv3543387 | Chr.8:39364918 - 39533981 on Build GRCh38 | Deletion |
|
| dgv1282e199 | Chr.8:39373726 - 39529923 on Build GRCh38 | Deletion |
|
| dgv12103n54 | Chr.8:39393511 - 39507504 on Build GRCh38 | Loss |
|
| dgv3807n106 | Chr.8:39374281 - 39529681 on Build GRCh38 | Deletion |
|
| nsv1024969 | Chr.8:39356415 - 39573619 on Build GRCh38 | Gain |
|
| nsv515081 | Chr.8:39375684 - 39528640 on Build GRCh38 | Loss |
|
| dgv147n17 | Chr.8:39444668 - 39547818 on Build GRCh38 | Loss |
|
| dgv12108n54 | Chr.8:39465868 - 39507199 on Build GRCh38 | Loss |
|
| nsv4435496 | Chr.8:39374556 - 39529709 on Build GRCh38 | Deletion |
|
| esv32816 | Chr.8:39374710 - 39531279 on Build GRCh38 | Loss |
|