Search
Search

Entrez Gene ID: | 3030 |
Gene Name: | hydroxyacyl-CoA dehydrogenase trifunctional multienzyme complex subunit alpha |
Gene Aliases: |
ECHA, GBP, HADH, LCEH, LCHAD, MLCL AT, MTPA, TP-ALPHA |
Location: |
Chr.2:26190635-26244632 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HADHA | NM_000182.5 | NP_000173.2 | ||
| AK293398.1 | BAG56906.1 | |||
| AK299258.1 | BAG61288.1 | |||
| AK302532.1 | BAG63804.1 | |||
| AK313027.1 | BAG35861.1 | |||
| BC009235.2 | AAH09235.1 | |||
| D16480.1 | BAA03941.1 | |||
| FJ626228.1 | ||||
| GU727642.1 | ADU87643.1 | |||
| U04627.1 | AAA56664.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33316 | Chr.2:25440017 - 26672800 on Build GRCh38 | Loss |
|
Set Membership: |
|