Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03512945_cn
          See other MAPK10 CNV Assays ›
          Gene Symbol
          MAPK10
          Assay Reference Genome
          Location

          Chr.4:86057438 on build GRCh38
          Cytoband
          4q21.3
          Assay ID Hs03512945_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5602

          Gene Name:

          mitogen-activated protein kinase 10

          Gene Aliases:

          JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK

          Location:

          Chr.4:86012296-86594117 on Build GRCh38

          Assay Gene Location:

          Within Intron 23
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MAPK10 NM_001318067.1 NP_001304996.1
          NM_001318068.1 NP_001304997.1
          NM_001318069.1 NP_001304998.1
          NM_002753.4 NP_002744.1
          NM_138980.3 NP_620446.1
          NM_138982.3 NP_620448.1
          XM_005263129.2 XP_005263186.1
          XM_005263130.2 XP_005263187.1
          XM_005263131.2 XP_005263188.1
          XM_005263135.3 XP_005263192.1
          XM_006714268.2 XP_006714331.1
          XM_006714269.2 XP_006714332.1
          XM_011532117.2 XP_011530419.1
          XM_011532118.2 XP_011530420.1
          XM_011532120.2 XP_011530422.1
          XM_011532121.2 XP_011530423.1
          XM_017008419.1 XP_016863908.1
          XM_017008420.1 XP_016863909.1
          XM_017008421.1 XP_016863910.1
          XM_017008422.1 XP_016863911.1
          XM_017008423.1 XP_016863912.1
          XM_017008424.1 XP_016863913.1
          XM_017008425.1 XP_016863914.1
          XM_017008426.1 XP_016863915.1
          XM_017008427.1 XP_016863916.1
          XM_017008428.1 XP_016863917.1
          XM_017008429.1 XP_016863918.1
          XM_017008430.1 XP_016863919.1
          XM_017008431.1 XP_016863920.1
          XM_017008432.1 XP_016863921.1
          XM_017008433.1 XP_016863922.1
          XM_017008434.1 XP_016863923.1
          XM_017008435.1 XP_016863924.1
          XM_017008436.1 XP_016863925.1
          XM_017008437.1 XP_016863926.1
          XM_017008438.1 XP_016863927.1
          XM_017008439.1 XP_016863928.1
          XM_017008440.1 XP_016863929.1
          XM_017008441.1 XP_016863930.1
          XM_017008442.1 XP_016863931.1
          XM_017008443.1 XP_016863932.1
          XM_017008444.1 XP_016863933.1
          XM_017008445.1 XP_016863934.1
          XM_017008446.1 XP_016863935.1
          XM_017008447.1 XP_016863936.1
          XM_017008448.1 XP_016863937.1
          XM_017008449.1 XP_016863938.1
          XM_017008450.1 XP_016863939.1
          XM_017008451.1 XP_016863940.1
          XM_017008452.1 XP_016863941.1
          AK057723.1
          AK091104.1
          AK124791.1
          AK225974.1
          AK294125.1
          BC035057.1 AAH35057.1
          BC051731.1 AAH51731.1
          BC065516.1 AAH65516.1
          U07620.1 AAC50101.1
          U34819.1 AAC50604.1
          U34820.1 AAC50605.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv10529 Chr.4:86054725 - 86059733 on Build GRCh38 Loss MAPK10
          nsv513143 Chr.4:86055189 - 86058905 on Build GRCh38 Loss MAPK10
          esv3601253 Chr.4:86055222 - 86058804 on Build GRCh38 Loss MAPK10
          nsv514230 Chr.4:86055343 - 86058719 on Build GRCh38 Loss MAPK10
          esv2727931 Chr.4:86055198 - 86058800 on Build GRCh38 Deletion MAPK10
          esv2678617 Chr.4:86055221 - 86058798 on Build GRCh38 Deletion MAPK10
          dgv5332n100 Chr.4:86044950 - 86062079 on Build GRCh38 Loss MAPK10
          esv2422128 Chr.4:86054941 - 86058791 on Build GRCh38 Deletion MAPK10
          nsv471766 Chr.4:86055234 - 86058799 on Build GRCh38 Loss MAPK10
          dgv9150n54 Chr.4:86057367 - 86058774 on Build GRCh38 Loss MAPK10
          esv3564185 Chr.4:86055085 - 86058922 on Build GRCh38 Deletion MAPK10
          nsv594760 Chr.4:86055260 - 86058774 on Build GRCh38 Loss MAPK10
          dgv1508e212 Chr.4:86054928 - 86058923 on Build GRCh38 Loss MAPK10
          dgv5334n100 Chr.4:86054929 - 86079056 on Build GRCh38 Loss MAPK10
          dgv5333n100 Chr.4:86053362 - 86070837 on Build GRCh38 Loss MAPK10
          nsv1003585 Chr.4:86036814 - 86329283 on Build GRCh38 Loss LOC101929064 MAPK10
          dgv9149n54 Chr.4:86056551 - 86058774 on Build GRCh38 Loss MAPK10
          dgv9147n54 Chr.4:86056124 - 86058774 on Build GRCh38 Loss MAPK10
          nsv511259 Chr.4:86045174 - 86062079 on Build GRCh38 Loss MAPK10
          esv26690 Chr.4:86055238 - 86058859 on Build GRCh38 Loss MAPK10
          nsv1161116 Chr.4:86054070 - 86058791 on Build GRCh38 Deletion MAPK10
          dgv2844n106 Chr.4:86055209 - 86058847 on Build GRCh38 Deletion MAPK10
          esv2763854 Chr.4:86053909 - 86062079 on Build GRCh38 Loss MAPK10

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          protein phosphorylation
          signal transduction
          JNK cascade
          JUN phosphorylation
          response to light stimulus
          Fc-epsilon receptor signaling pathway
          regulation of circadian rhythm
          rhythmic process
          regulation of sequence-specific DNA binding transcription factor activity
          JUN kinase activity
          MAP kinase kinase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline