Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03586990_cn
          See other PSORS1C3 CNV Assays ›
          Gene Symbol
          PSORS1C3
          Assay Reference Genome
          Location

          Chr.6:31175519 on build GRCh38
          Cytoband
          Assay ID Hs03586990_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          100130889

          Gene Name:

          psoriasis susceptibility 1 candidate 3

          Gene Aliases:

          NCRNA00196

          Location:

          Chr.6:2656176-2668769 on Build GRCh38

          Assay Gene Location:

          Within Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PSORS1C3 NR_026816.2
          NR_152828.1
          NR_152829.1
          NR_152830.1
          NR_152831.1
          NR_152832.1
          NR_152833.1
          NR_152834.1
          NR_152835.1
          NR_152836.1
          NR_152837.1
          NR_152838.1
          NR_152839.1
          AB932952.1
          AB932953.1
          AB932954.1
          AB933312.1
          AB933313.1
          AB933320.1
          AB933321.1
          AY484516.1 AAS49640.1
          LC027944.1
          LC027945.1
          LC027946.1
          LC050979.1
          LC050980.1
          LC050981.1
          LC050982.1
          LC050983.1
          LC050984.1
          LC050985.1
          LC050986.1
          LC050987.1
          LC050988.1
          LC050989.1
          LC050990.1
          LC050991.1
          OA988594.1 1 1073
          OA988595.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2759414 Chr.6:31060513 - 31574531 on Build GRCh38 Gain+Loss POU5F1 SNORD117 PSORS1C3 PSORS1C2 ATP6V1G2 MICA-AS1 DDX39B ATP6V1G2-DDX39B LINC01149 MCCD1 PSORS1C1 MICA TCF19 SNORD84 CDSN HCP5 MICB-DT LINC02571 HCG26 HCG27 LTA DDX39B-AS1 HLA-B C6orf15 NFKBIL1 CCHCR1 HLA-C LOC100287329 MIR6891 MICB
          esv2731791 Chr.6:30404802 - 31346410 on Build GRCh38 Deletion POU5F1 PSORS1C3 PSORS1C2 ABCF1 DDR1 MUC21 MUC22 MUCL3 HCG21 C6orf136 HLA-E IER3-AS1 MDC1-AS1 PSORS1C1 TUBB MRPS18B VARS2 GNL1 SFTA2 GTF2H4 IER3 TCF19 ATAT1 MIR877 LINC00243 HCG20 CDSN DHX16 LINC02569 LINC02570 LINC02571 NRM LOC105375012 HCG27 MDC1 HCG22 LOC124901299 C6orf15 PRR3 PPP1R10 CCHCR1 MIR4640 PPP1R18 HLA-C FLOT1
          dgv1174e214 Chr.6:31139843 - 31176217 on Build GRCh38 Gain POU5F1 PSORS1C1 PSORS1C3 CCHCR1 TCF19
          esv3608532 Chr.6:31163674 - 31304530 on Build GRCh38 Gain HCG27 POU5F1 PSORS1C3 LINC02571 HLA-C TCF19
          dgv1175e214 Chr.6:31170668 - 31187842 on Build GRCh38 Gain POU5F1 PSORS1C3
          nsv1074389 Chr.6:29116522 - 31446623 on Build GRCh38 Deletion TRIM10 PSORS1C3 SNORD32B MUC21 OR2I1 HCG21 HLA-E IER3-AS1 TUBB HLA-H HLA-J HCP5B HLA-F HCG20 LINC02569 LINC01015 OR11A1 HCG9 NRM IFITM4P LOC124901299 TRIM15 OR2H1 POLR1H OR12D2 ABCF1 MUC22 MICA-AS1 OR2H2 TRIM26 MDC1-AS1 MICA SFTA2 GTF2H4 IER3 RNF39 ATAT1 MIR877 LINC00243 TRIM40 CDSN DHX16 TRIM31 TRIM31-AS1 MAS1L LOC105375012 POLR1HASP HCG22 PRR3 CCHCR1 MOG MIR6891 POU5F1 PSORS1C2 HLA-G HCG17 MUCL3 HLA-V LINC01149 MRPS18B OR5V1 HLA-L RPP21 LINC02570 HCG27 LINC03003 HLA-B PPP1R10 MIR4640 PPP1R18 TRIM39 FLOT1 OR2J2 OR12D3 DDR1 HLA-A LINC02829 C6orf136 OR10C1 HLA-K PSORS1C1 HCG18 VARS2 GNL1 OR14J1 HLA-F-AS1 TCF19 OR12D1 GABBR1 TRIM39-RPP21 HCG4B LINC02571 ZFP57 MDC1 PPP1R11 C6orf15 HLA-C UBD HCG4
          dgv1019n145 Chr.6:29873402 - 32666862 on Build GRCh38 Deletion TRIM10 SNHG32 SNORD117 PSORS1C3 C6orf47 MUC21 LTB TNF HLA-DQB1-AS1 HCG21 HLA-E MCCD1 IER3-AS1 LY6G6C LY6G6D TUBB C4A NOTCH4 AGER LY6G6F HSPA1L HLA-H PRRC2A HLA-J HCP5B NEU1 DXO BTNL2 C4B HCG20 HCP5 LINC02569 HCG9 LY6G6E NRM EHMT2-AS1 SAPCD1 LOC124901299 TNXA TRIM15 APOM POLR1H BAG6 CFB GPANK1 LY6G5B ABCF1 MUC22 MICA-AS1 HLA-DRB5 VARS1 LST1 FKBPL CYP21A1P TRIM26 MDC1-AS1 TNXB HSPA1A MICA SFTA2 GTF2H4 IER3 RNF39 LSM2 ATAT1 MIR877 C2 SNORD48 LINC00243 TRIM40 CDSN DHX16 TRIM31 HLA-DQA1 PRRT1 MIR6721 GPSM3 TRIM31-AS1 ZBTB12 LOC105375012 MSH5-SAPCD1 NCR3 AIF1 POLR1HASP HCG22 HLA-DRB1 PRR3 CCHCR1 TSBP1 MIR1236 LOC100287329 MIR6891 WHR1 POU5F1 PSORS1C2 SNORA38 HCG17 MUCL3 ATP6V1G2-DDX39B LINC01149 SKIC2 MRPS18B MPIG6B ABHD16A CSNK2B HLA-L RPP21 C2-AS1 MICB-DT LINC02570 MIR6833 NELFE HCG26 SAPCD1-AS1 HCG27 LY6G6F-LY6G6D HLA-DRA DDX39B-AS1 HLA-B PPP1R10 DDAH2 RNF5 MIR4640 PPP1R18 TRIM39 FLOT1 MICB LOC100507547 CYP21A2 MIR4646 PBX2 DDR1 ATP6V1G2 HLA-A SNORD52 DDX39B MIR6832 C6orf136 VWA7 HLA-K LY6G5C CLIC1 TSBP1-AS1 PSORS1C1 HCG18 HCG23 EHMT2 VARS2 GNL1 PPT2 TCF19 HLA-DRB6 SNORD84 TRIM39-RPP21 ATF6B HCG4B LINC02571 EGFL8 HLA-DQB1 PPT2-EGFL8 LTA AGPAT1 MDC1 HSPA1B PPP1R11 C6orf15 NFKBIL1 HLA-C SLC44A4 MSH5
          nsv601481 Chr.6:30454855 - 31316476 on Build GRCh38 Loss POU5F1 PSORS1C3 PSORS1C2 ABCF1 DDR1 MUC21 MUC22 MUCL3 HCG21 C6orf136 HLA-E IER3-AS1 MDC1-AS1 PSORS1C1 TUBB MRPS18B VARS2 GNL1 SFTA2 GTF2H4 IER3 TCF19 ATAT1 MIR877 LINC00243 HCG20 CDSN DHX16 LINC02569 LINC02570 LINC02571 NRM LOC105375012 HCG27 MDC1 HCG22 LOC124901299 C6orf15 PRR3 PPP1R10 CCHCR1 MIR4640 PPP1R18 HLA-C FLOT1
          nsv1122126 Chr.6:28628023 - 31244023 on Build GRCh38 Deletion TRIM10 PSORS1C3 SNORD32B MUC21 TRIM27 OR2I1 HCG21 HLA-E IER3-AS1 TUBB ZNF311 HLA-H OR2J3 HLA-J HCP5B HLA-F HCG20 LINC02569 LINC01015 OR11A1 HCG9 NRM OR2W1-AS1 IFITM4P LINC01556 LOC124901299 TRIM15 OR2H1 POLR1H OR12D2 ABCF1 MUC22 OR2H2 TRIM26 MDC1-AS1 SFTA2 GTF2H4 IER3 RNF39 ATAT1 MIR877 LINC00243 TRIM40 CDSN DHX16 TRIM31 TRIM31-AS1 MAS1L LOC105375012 POLR1HASP HCG22 PRR3 CCHCR1 MOG LINC00533 POU5F1 PSORS1C2 HLA-G HCG17 MUCL3 HLA-V HCG14 MRPS18B OR2W1 HCG15 OR5V1 HLA-L RPP21 LINC02570 HCG27 LINC03003 OR2B3 PPP1R10 MIR4640 PPP1R18 TRIM39 FLOT1 OR2J2 OR12D3 DDR1 HLA-A LINC02829 C6orf136 OR10C1 HLA-K PSORS1C1 HCG18 VARS2 GNL1 OR14J1 HLA-F-AS1 TCF19 OR12D1 GABBR1 TRIM39-RPP21 HCG4B LINC01623 ZFP57 MDC1 PPP1R11 OR2J1 C6orf15 UBD HCG4
          nsv1144137 Chr.6:29720323 - 31338123 on Build GRCh38 Deletion TRIM10 POU5F1 PSORS1C3 PSORS1C2 MUC21 HLA-G HCG17 MUCL3 HCG21 HLA-E HLA-V IER3-AS1 TUBB MRPS18B HLA-H HLA-J HCP5B HLA-F HLA-L HCG20 RPP21 LINC02569 LINC02570 HCG9 NRM IFITM4P HCG27 LOC124901299 PPP1R10 MIR4640 TRIM15 PPP1R18 TRIM39 POLR1H FLOT1 ABCF1 DDR1 MUC22 HLA-A C6orf136 TRIM26 HLA-K MDC1-AS1 PSORS1C1 HCG18 VARS2 GNL1 SFTA2 GTF2H4 IER3 RNF39 HLA-F-AS1 TCF19 ATAT1 MIR877 TRIM39-RPP21 LINC00243 TRIM40 CDSN DHX16 TRIM31 HCG4B LINC02571 TRIM31-AS1 LOC105375012 MDC1 PPP1R11 POLR1HASP HCG22 C6orf15 PRR3 CCHCR1 HLA-C HCG4
          nsv601264 Chr.6:29883666 - 31320716 on Build GRCh38 Loss TRIM10 POU5F1 PSORS1C3 PSORS1C2 MUC21 HCG17 MUCL3 HCG21 HLA-E IER3-AS1 TUBB MRPS18B HLA-H HLA-J HLA-L HCG20 RPP21 LINC02569 LINC02570 HCG9 NRM HCG27 LOC124901299 PPP1R10 MIR4640 TRIM15 PPP1R18 TRIM39 POLR1H FLOT1 ABCF1 DDR1 MUC22 HLA-A C6orf136 TRIM26 HLA-K MDC1-AS1 PSORS1C1 HCG18 VARS2 GNL1 SFTA2 GTF2H4 IER3 RNF39 TCF19 ATAT1 MIR877 TRIM39-RPP21 LINC00243 TRIM40 CDSN DHX16 TRIM31 HCG4B LINC02571 TRIM31-AS1 LOC105375012 MDC1 PPP1R11 POLR1HASP HCG22 C6orf15 PRR3 CCHCR1 HLA-C
          esv32635 Chr.6:31173727 - 31282841 on Build GRCh38 Gain+Loss HCG27 PSORS1C3 HLA-C
          esv3411275 Chr.6:29981607 - 31490616 on Build GRCh38 Duplication TRIM10 POU5F1 PSORS1C3 PSORS1C2 MUC21 HCG17 MUCL3 HCG21 HLA-E LINC01149 IER3-AS1 TUBB MRPS18B HLA-J HLA-L HCG20 RPP21 HCP5 LINC02569 MICB-DT LINC02570 NRM HCG26 HCG27 HLA-B LOC124901299 PPP1R10 MIR4640 TRIM15 PPP1R18 TRIM39 POLR1H FLOT1 ABCF1 DDR1 MUC22 MICA-AS1 C6orf136 TRIM26 MDC1-AS1 PSORS1C1 HCG18 VARS2 GNL1 MICA SFTA2 GTF2H4 IER3 RNF39 TCF19 ATAT1 MIR877 TRIM39-RPP21 LINC00243 TRIM40 CDSN DHX16 TRIM31 LINC02571 TRIM31-AS1 LOC105375012 MDC1 PPP1R11 POLR1HASP HCG22 C6orf15 PRR3 CCHCR1 HLA-C MIR6891
          nsv5460471 Chr.6:31119831 - 31186318 on Build GRCh38 Duplication CDSN POU5F1 PSORS1C1 PSORS1C3 PSORS1C2 CCHCR1 TCF19

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline