Search
Search

Entrez Gene ID: | 7410 |
Gene Name: | vav guanine nucleotide exchange factor 2 |
Gene Aliases: |
VAV-2 |
Location: |
Chr.9:133761894-133992374 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VAV2 | NM_001134398.1 | NP_001127870.1 | ||
| NM_003371.3 | NP_003362.2 | |||
| XM_005272213.1 | XP_005272270.1 | |||
| XM_017015108.1 | XP_016870597.1 | |||
| XM_017015109.1 | XP_016870598.1 | |||
| XM_017015110.1 | XP_016870599.1 | |||
| XM_017015111.1 | XP_016870600.1 | |||
| XM_017015112.1 | XP_016870601.1 | |||
| XM_017015113.1 | XP_016870602.1 | |||
| BC132965.1 | ||||
| BC132967.1 | ||||
| BQ050947.1 | ||||
| BX640754.1 | CAE45861.1 | |||
| S76992.1 | AAB34377.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| nsv951802 | Chr.9:133753679 - 133815978 on Build GRCh38 | Deletion |
|
| dgv95n68 | Chr.9:133728580 - 133915151 on Build GRCh38 | Loss |
|
| nsv482984 | Chr.9:133676624 - 133855751 on Build GRCh38 | Gain |
|
Set Membership: |
|