Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03746773_cn
          See other AKR1C1 CNV Assays ›
          Gene Symbol
          AKR1C1
          Assay Reference Genome
          Location

          Chr.10:4966466 on build GRCh38
          Cytoband
          Assay ID Hs03746773_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1645

          Gene Name:

          aldo-keto reductase family 1 member C1

          Gene Aliases:

          2-ALPHA-HSD, 20-ALPHA-HSD, C9, DD1, DD1/DD2, DDH, DDH1, H-37, HAKRC, HBAB, MBAB

          Location:

          Chr.10:4963415-4983283 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          AKR1C1 NM_001353.6 NP_001344.2
          AB031083.1 BAA92883.1
          AK303435.1 BAG64483.1
          AK310658.1
          BC015490.2 AAH15490.1
          BC020216.1 AAH20216.1
          BC040210.1 AAH40210.1
          BM923712.1
          BT007197.1 AAP35861.1
          D26124.1 BAA05121.1
          M86609.1 AAB02880.1
          S68290.1 AAD14012.1
          U05684.1 AAA16227.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2763913 Chr.10:4918829 - 5279444 on Build GRCh38 Gain+Loss LOC101928051 AKR1C4 LINC02561 AKR1C1 AKR1C8 AKR1C3 AKR1C2
          nsv549882 Chr.10:4901326 - 5161901 on Build GRCh38 Loss LOC101928051 AKR1C1 AKR1C8 AKR1C3 AKR1C2 AKR1C6P
          nsv4184242 Chr.10:4907689 - 4993341 on Build GRCh38 Deletion AKR1C1 AKR1C2 AKR1C6P
          nsv3238849 Chr.10:4957420 - 5009066 on Build GRCh38 Insertion LOC101928051 AKR1C1 AKR1C2
          nsv947726 Chr.10:4965877 - 4969892 on Build GRCh38 Duplication AKR1C1
          nsv1039386 Chr.10:4966191 - 5344882 on Build GRCh38 Gain LOC101928051 AKR1C4 LINC02561 AKR1C1 AKR1C8 AKR1C3 AKR1C2
          nsv4185641 Chr.10:4942308 - 5006308 on Build GRCh38 Duplication LOC101928051 AKR1C1 AKR1C2
          nsv825221 Chr.10:4957157 - 5004047 on Build GRCh38 Loss LOC101928051 AKR1C1 AKR1C2
          nsv4526555 Chr.10:4884807 - 5206537 on Build GRCh38 Duplication LOC101928051 AKR1C4 AKR1C1 AKR1C8 AKR1C3 AKR1C2 AKR1C6P
          dgv601n223 Chr.10:4827838 - 5282828 on Build GRCh38 Deletion LOC101928051 AKR1C4 LINC02561 AKR1C1 AKR1C8 AKR1E2 AKR1C3 AKR1C2 AKR1C6P
          nsv549881 Chr.10:4859606 - 5277716 on Build GRCh38 Loss LOC101928051 AKR1C4 LINC02561 AKR1C1 AKR1C8 AKR1E2 AKR1C3 AKR1C2 AKR1C6P
          nsv6014032 Chr.10:4929820 - 5080261 on Build GRCh38 Deletion LOC101928051 AKR1C1 AKR1C3 AKR1C2
          nsv1045155 Chr.10:4830047 - 5169355 on Build GRCh38 Loss LOC101928051 AKR1C1 AKR1C8 AKR1E2 AKR1C3 AKR1C2 AKR1C6P
          esv3578596 Chr.10:4909566 - 5001136 on Build GRCh38 Loss LOC101928051 AKR1C1 AKR1C2 AKR1C6P
          nsv1050246 Chr.10:4885236 - 5169355 on Build GRCh38 Gain LOC101928051 AKR1C1 AKR1C8 AKR1C3 AKR1C2 AKR1C6P
          nsv6454243 Chr.10:4952198 - 4970252 on Build GRCh38 Deletion AKR1C1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          alcohol metabolic process
          lipid metabolic process
          prostaglandin metabolic process
          xenobiotic metabolic process
          digestion
          steroid metabolic process
          bile acid metabolic process
          bile acid and bile salt transport
          intestinal cholesterol absorption
          epithelial cell differentiation
          monocarboxylic acid metabolic process
          hormone metabolic process
          progesterone metabolic process
          retinal metabolic process
          cholesterol homeostasis
          daunorubicin metabolic process
          doxorubicin metabolic process
          cellular response to jasmonic acid stimulus
          positive regulation of reactive oxygen species metabolic process
          alditol:NADP+ 1-oxidoreductase activity
          estradiol 17-beta-dehydrogenase activity
          protein binding
          alcohol dehydrogenase (NADP+) activity
          oxidoreductase activity
          oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
          testosterone dehydrogenase [NAD(P)] activity
          carboxylic acid binding
          bile acid binding
          3-beta-hydroxy-5-beta-steroid dehydrogenase (NADP+) activity
          steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          17-alpha,20-alpha-dihydroxypregn-4-en-3-one dehydrogenase activity
          androsterone dehydrogenase activity
          5-alpha-androstane-3-beta,17-beta-diol dehydrogenase (NADP+) activity
          androstan-3-alpha,17-beta-diol dehydrogenase activity
          testosterone 17-beta-dehydrogenase (NADP+) activity
          ketosteroid monooxygenase activity
          trans-1,2-dihydrobenzene-1,2-diol dehydrogenase activity
          indanol dehydrogenase activity
          3-alpha-hydroxysteroid 3-dehydrogenase [NAD(P)+] activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline