Search
Search

Entrez Gene ID: | 5214 |
Gene Name: | phosphofructokinase, platelet |
Gene Aliases: |
ATP-PFK, PFK-C, PFK-P, PFKF |
Location: |
Chr.10:3067548-3136805 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549814 | Chr.10:3055020 - 3170233 on Build GRCh38 | Loss |
|
| esv3622165 | Chr.10:3099237 - 3125437 on Build GRCh38 | Gain |
|
| dgv932n54 | Chr.10:2382612 - 3253481 on Build GRCh38 | Gain |
|
| nsv428229 | Chr.10:2618610 - 3156581 on Build GRCh38 | Gain |
|
| esv2730995 | Chr.10:2770220 - 3706473 on Build GRCh38 | Deletion |
|
| nsv6442021 | Chr.10:3030896 - 3173781 on Build GRCh38 | Duplication |
|
| esv2750891 | Chr.10:3098894 - 3164173 on Build GRCh38 | Loss |
|
| nsv470913 | Chr.10:3052993 - 3125271 on Build GRCh38 | Loss |
|
| nsv6442520 | Chr.10:2846098 - 3581354 on Build GRCh38 | Duplication |
|
| nsv549810 | Chr.10:3035272 - 3691194 on Build GRCh38 | Gain |
|
| esv28525 | Chr.10:3120877 - 3123679 on Build GRCh38 | Loss |
|
| nsv6454219 | Chr.10:2905224 - 3284256 on Build GRCh38 | Duplication |
|
| nsv6437272 | Chr.10:3058401 - 3259500 on Build GRCh38 | Deletion |
|
| nsv549818 | Chr.10:3081241 - 3227125 on Build GRCh38 | Gain |
|
| nsv4179484 | Chr.10:2460844 - 3257750 on Build GRCh38 | Duplication |
|
| dgv935n54 | Chr.10:3044294 - 3125271 on Build GRCh38 | Loss |
|
| dgv38n206 | Chr.10:2613782 - 3185857 on Build GRCh38 | Duplication |
|
| nsv6455282 | Chr.10:3085901 - 3154200 on Build GRCh38 | Deletion |
|
Set Membership: |
|