Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03757892_cn
          See other DMBT1 CNV Assays ›
          Gene Symbol
          DMBT1
          Assay Reference Genome
          Location

          Chr.10:122641444 on build GRCh38
          Cytoband
          10q26.13
          Assay ID Hs03757892_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1755

          Gene Name:

          deleted in malignant brain tumors 1

          Gene Aliases:

          GP340, SAG, muclin

          Location:

          Chr.10:122560665-122643736 on Build GRCh38

          Assay Gene Location:

          Within Intron 56
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          DMBT1 NM_001320644.1 NP_001307573.1
          NM_004406.2 NP_004397.2
          NM_007329.2 NP_015568.2
          NM_017579.2 NP_060049.2
          XM_006717660.3 XP_006717723.1
          XM_006717665.3 XP_006717728.1
          XM_011539388.2 XP_011537690.1
          XM_011539389.2 XP_011537691.1
          XM_011539390.2 XP_011537692.1
          XM_011539391.2 XP_011537693.1
          XM_011539392.2 XP_011537694.1
          XM_011539393.2 XP_011537695.1
          XM_011539394.2 XP_011537696.1
          XM_011539395.2 XP_011537697.1
          XM_011539396.2 XP_011537698.1
          XM_011539398.2 XP_011537700.1
          XM_011539399.2 XP_011537701.1
          XM_011539400.2 XP_011537702.1
          XM_011539401.2 XP_011537703.1
          XM_011539402.2 XP_011537704.1
          XM_011539403.2 XP_011537705.1
          XM_011539405.2 XP_011537707.1
          XM_011539407.2 XP_011537709.1
          XM_011539408.2 XP_011537710.1
          XM_011539409.2 XP_011537711.1
          XM_011539410.2 XP_011537712.1
          XM_011539411.2 XP_011537713.1
          XM_011539413.2 XP_011537715.1
          XM_011539414.2 XP_011537716.1
          XM_011539415.2 XP_011537717.1
          XM_017015798.1 XP_016871287.1
          AF159456.1 AAD49696.1
          AJ000342.1 CAA04019.1
          AJ243212.1 CAB56155.1
          AJ243224.1 CAB63942.1
          AJ297935.1 CAC44122.1
          AK304149.1
          BC153299.1
          BX640988.1 CAE45995.1
          FJ234410.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv428240 Chr.10:122503559 - 122695299 on Build GRCh38 Gain+Loss HTRA1 DMBT1

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs75964629] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          pattern recognition receptor signaling pathway
          receptor-mediated endocytosis
          multicellular organism development
          protein transport
          epithelial cell differentiation
          induction of bacterial agglutination
          cellular protein metabolic process
          innate immune response
          defense response to virus
          scavenger receptor activity
          protein binding
          signaling pattern recognition receptor activity
          zymogen binding
          calcium-dependent protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline