Search
Search

Entrez Gene ID: | 338653 |
Gene Name: | KCNQ1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:2840135-2861569 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ1-AS1 | NR_130721.1 | |||
| AJ251642.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759795 | Chr.11:2767814 - 2942382 on Build GRCh38 | Gain |
|
| nsv951292 | Chr.11:2792271 - 2927870 on Build GRCh38 | Deletion |
|
| nsv825715 | Chr.11:2855771 - 2890367 on Build GRCh38 | Gain |
|
| nsv469930 | Chr.11:2831299 - 2924715 on Build GRCh38 | Loss |
|
| nsv1053066 | Chr.11:2829552 - 2872696 on Build GRCh38 | Gain |
|
| nsv553111 | Chr.11:2842057 - 2904391 on Build GRCh38 | Loss |
|
| dgv1574n54 | Chr.11:2848935 - 2889391 on Build GRCh38 | Loss |
|