Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03792881_cn
          See other MRPL23 CNV Assays ›
          Gene Symbol
          MRPL23
          Assay Reference Genome
          Location

          Chr.11:2011148 on build GRCh38
          Cytoband
          Assay ID Hs03792881_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          6150

          Gene Name:

          mitochondrial ribosomal protein L23

          Gene Aliases:

          L23MRP, RPL23, RPL23L, uL23m

          Location:

          Chr.11:139854-207466 on Build GRCh38

          Assay Gene Location:

          Within Intron 9
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MRPL23 NM_001400176.1 NP_001387105.1
          XM_011520273.2 XP_011518575.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1047474 Chr.11:1803515 - 2043381 on Build GRCh38 Gain PRR33 MRPL23-AS1 SYT8 TNNI2 H19 SNORD131 MIR4298 LINC01219 MIR7847 TNNT3 MRPL23 MIR675 LSP1 LINC01150
          dgv1016n100 Chr.11:1946041 - 2043381 on Build GRCh38 Gain MRPL23-AS1 H19 SNORD131 MRPL23 MIR675 LINC01219
          nsv553065 Chr.11:2011014 - 2036417 on Build GRCh38 Loss MRPL23
          nsv469926 Chr.11:1813024 - 2036416 on Build GRCh38 Loss PRR33 MRPL23-AS1 SYT8 TNNI2 H19 SNORD131 MIR4298 LINC01219 MIR7847 TNNT3 MRPL23 MIR675 LSP1 LINC01150
          nsv6144247 Chr.11:1983000 - 2041000 on Build GRCh38 Deletion MRPL23-AS1 H19 MRPL23 MIR675 LINC01219
          nsv951283 Chr.11:1913571 - 2040070 on Build GRCh38 Deletion PRR33 MRPL23-AS1 H19 TNNT3 SNORD131 MRPL23 MIR675 LINC01219
          esv29980 Chr.11:1820597 - 2097889 on Build GRCh38 Loss PRR33 MRPL23-AS1 SYT8 TNNI2 H19 SNORD131 MIR4298 LINC01219 MIR7847 TNNT3 MRPL23 MIR675 LSP1 LINC01150
          nsv467645 Chr.11:1733170 - 2418537 on Build GRCh38 Gain MRPL23-AS1 TNNI2 H19 SNORD131 CD81-AS1 CD81 TRPM5 TSSC4 LINC01219 MIR483 TNNT3 MRPL23 LSP1 LINC01150 PRR33 TSPAN32 SYT8 INS INS-IGF2 IGF2-AS CTSD MIR4298 PRADX TH IGF2 MIR7847 IFITM10 MIR4686 MIR675 ASCL2 C11orf21

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          mitochondrial translation
          RNA binding
          structural constituent of ribosome
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline