Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03796244_cn
          See other EEF1G CNV Assays ›
          Gene Symbol
          EEF1G
          Assay Reference Genome
          Location

          Chr.11:62570432 on build GRCh38
          Cytoband
          11q12.3
          Assay ID Hs03796244_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1937

          Gene Name:

          eukaryotic translation elongation factor 1 gamma

          Gene Aliases:

          EF1G, GIG35

          Location:

          Chr.11:62559601-62573988 on Build GRCh38

          Assay Gene Location:

          Within Intron 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          EEF1G NM_001404.4 NP_001395.1
          AK092787.1
          AK129569.1
          AK129618.1
          AK130026.1
          AK299876.1
          AK300203.1
          AK300686.1
          AK300729.1
          AL556034.3
          AY542307.1 AAT08176.1
          AY687925.1
          BQ947806.1
          M55409.1 AAC18414.1
          X63526.1 CAA45089.1
          Z11531.1 CAA77630.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3584599 Chr.11:62495270 - 62630578 on Build GRCh38 Gain ROM1 MTA2 EEF1G MIR6747 AHNAK TUT1 EML3 B3GAT3 GANAB
          nsv832180 Chr.11:62434133 - 62597356 on Build GRCh38 Loss MTA2 EEF1G MIR6747 AHNAK TUT1
          nsv347 Chr.11:62529596 - 62571455 on Build GRCh38 Deletion EEF1G MIR6747 AHNAK
          nsv825939 Chr.11:62558611 - 62643105 on Build GRCh38 Gain ROM1 MTA2 EEF1G MIR6747 TUT1 EML3 B3GAT3 GANAB
          nsv832182 Chr.11:62528103 - 62695376 on Build GRCh38 Gain+Loss ROM1 MTA2 UQCC3 EEF1G SNORA57 MIR6747 LRRN4CL METTL12 EML3 GANAB HNRNPUL2-BSCL2 BSCL2 INTS5 AHNAK C11orf98 TUT1 B3GAT3 UBXN1 LBHD1
          nsv555174 Chr.11:62566624 - 62729047 on Build GRCh38 Gain ROM1 MTA2 UQCC3 EEF1G SNORA57 MIR6747 LRRN4CL METTL12 GNG3 EML3 GANAB HNRNPUL2-BSCL2 BSCL2 INTS5 HNRNPUL2 TTC9C C11orf98 TUT1 B3GAT3 UBXN1 LBHD1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          glutathione metabolic process
          response to virus
          cell-cell adhesion
          translation elongation factor activity
          glutathione transferase activity
          protein binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline