Search
Search

Entrez Gene ID: | 51564 |
Gene Name: | histone deacetylase 7 |
Gene Aliases: |
HD7, HD7A, HDAC7A |
Location: |
Chr.12:47782722-47821344 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC7 | NM_001098416.4 | NP_001091886.1 | ||
| NM_001308090.2 | NP_001295019.1 | |||
| NM_001368046.1 | NP_001354975.1 | |||
| NM_015401.5 | NP_056216.2 | |||
| NR_160435.1 | ||||
| NR_160436.1 | ||||
| XM_011538481.2 | XP_011536783.2 | |||
| XM_024449018.2 | 1 | 360 | XP_024304786.1 | |
| XM_047428978.1 | XP_047284934.1 | |||
| XM_047428979.1 | XP_047284935.1 | |||
| XM_047428980.1 | XP_047284936.1 | |||
| XM_047428981.1 | 1 | 359 | XP_047284937.1 | |
| XM_047428983.1 | XP_047284939.1 | |||
| AK225910.1 | ||||
| AK299292.1 | BAG61307.1 | |||
| AK303481.1 | BAG64517.1 | |||
| AY302468.1 | AAQ18232.1 | |||
| AY321367.1 | AAP84704.1 | |||
| BC064840.1 | AAH64840.1 | |||
| DC376749.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv94n21 | Chr.12:47725779 - 47821397 on Build GRCh38 | Loss |
|
| esv3693398 | Chr.12:47792955 - 47821966 on Build GRCh38 | Loss |
|
| esv999028 | Chr.12:47805518 - 47815349 on Build GRCh38 | Deletion |
|