Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03867191_cn
          See other CRIP2 CNV Assays ›
          Gene Symbol
          CRIP2
          Assay Reference Genome
          Location

          Chr.14:105477579 on build GRCh38
          Cytoband
          14q32.33
          Assay ID Hs03867191_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1397

          Gene Name:

          cysteine rich protein 2

          Gene Aliases:

          CRIP, CRP2, ESP1

          Location:

          Chr.14:105472938-105480170 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 4 - Intron 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CRIP2 NM_001270837.1 NP_001257766.1
          NM_001270841.1 NP_001257770.1
          NM_001312.3 NP_001303.1
          NR_073081.1
          NR_073082.1
          NR_073083.1
          NR_073084.1
          NR_073085.1
          AK057837.1
          AK091845.1
          AK223251.1 BAD96971.1
          AK300092.1
          AK311402.1
          AK315757.1
          BC000434.2 AAH00434.1
          BC001931.1 AAH01931.1
          BC034151.1 AAH34151.1
          BC128101.1
          BE271979.1
          BT019911.1 AAV38714.1
          CN367966.1
          D42123.1 BAA07703.1
          HY078632.1
          HY161884.1
          U36190.1 AAB03194.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv3996n54 Chr.14:105474226 - 105488519 on Build GRCh38 Gain CRIP1 CRIP2
          nsv566204 Chr.14:105376842 - 105479048 on Build GRCh38 Gain PACS2 LOC100507437 CRIP2 TEX22 MTA1
          nsv817647 Chr.14:105382554 - 105480505 on Build GRCh38 Gain PACS2 LOC100507437 CRIP2 TEX22 MTA1
          dgv3998n54 Chr.14:105474495 - 105770768 on Build GRCh38 Loss IGHA2 CRIP1 IGH MIR8071-1 CRIP2 LOC107987211 IGHG4 IGHG1 TMEM121 LOC105370697 IGHG2 C14orf80 IGHA1 IGHG3 MIR8071-2 ELK2AP
          nsv1160204 Chr.14:105291411 - 105483783 on Build GRCh38 Duplication PACS2 LOC100507437 CRIP2 BRF1 TEX22 MTA1
          dgv3994n54 Chr.14:105474172 - 105477662 on Build GRCh38 Gain CRIP2
          nsv566206 Chr.14:105439736 - 105543235 on Build GRCh38 Loss CRIP1 LOC105370697 CRIP2 C14orf80 MTA1 TMEM121
          nsv832890 Chr.14:105304955 - 105483089 on Build GRCh38 Loss PACS2 LOC100507437 CRIP2 BRF1 TEX22 MTA1
          nsv1440 Chr.14:105473170 - 105479862 on Build GRCh38 Insertion CRIP2
          esv1004211 Chr.14:105477270 - 105479324 on Build GRCh38 Insertion CRIP2
          nsv566210 Chr.14:105465248 - 105488368 on Build GRCh38 Gain CRIP1 CRIP2 MTA1
          nsv9177 Chr.14:105476516 - 105485998 on Build GRCh38 Gain CRIP2
          nsv827088 Chr.14:105454469 - 105498549 on Build GRCh38 Gain CRIP1 CRIP2 C14orf80 MTA1
          nsv952282 Chr.14:105363764 - 105510163 on Build GRCh38 Deletion CRIP1 PACS2 LOC100507437 CRIP2 C14orf80 TEX22 MTA1
          dgv3999n54 Chr.14:105474867 - 105487998 on Build GRCh38 Loss CRIP1 CRIP2
          esv2760014 Chr.14:105047009 - 105483089 on Build GRCh38 Loss GPR132 JAG2 PACS2 BTBD6 LOC100507437 LOC102723354 CRIP2 BRF1 MIR6765 NUDT14 TEX22 MTA1
          dgv3988n54 Chr.14:105466757 - 105477662 on Build GRCh38 Gain CRIP2 MTA1
          nsv470679 Chr.14:105124547 - 105605978 on Build GRCh38 Loss IGHA2 CRIP1 IGH BTBD6 CRIP2 BRF1 TEX22 TMEM121 JAG2 PACS2 LOC105370697 LOC100507437 C14orf80 MIR6765 NUDT14 MTA1
          dgv3987n54 Chr.14:105461599 - 105499970 on Build GRCh38 Loss CRIP1 CRIP2 C14orf80 MTA1
          nsv566265 Chr.14:105474766 - 105479048 on Build GRCh38 Loss CRIP2
          nsv1054190 Chr.14:105246882 - 105701138 on Build GRCh38 Gain IGHA2 CRIP1 IGH BTBD6 MIR8071-1 CRIP2 BRF1 IGHG4 TEX22 TMEM121 PACS2 LOC105370697 LOC100507437 IGHG2 C14orf80 MIR8071-2 ELK2AP MTA1
          esv3635645 Chr.14:105473682 - 105484540 on Build GRCh38 Gain CRIP2
          nsv517055 Chr.14:105136282 - 106779068 on Build GRCh38 Gain+Loss IGHA2 IGHD CRIP1 IGH BTBD6 BRF1 MIR4539 MIR4507 LOC107987211 IGHG4 IGHG1 MIR4537 IGHV3-41 IGHV4-31 LINC00221 PACS2 LOC105370697 IGHG2 IGHA1 MIR8071-2 LINC00226 MTA1 IGHV3-23 IGHV3-48 MIR8071-1 KIAA0125 IGHV3-30 CRIP2 ADAM6 TEX22 IGHV4-55 TMEM121 JAG2 IGHM LOC100507437 C14orf80 MIR6765 IGHV3-69-1 NUDT14 MIR4538 IGHG3 IGHV3-71 ELK2AP IGHV1-69
          dgv3991n54 Chr.14:105472502 - 105480505 on Build GRCh38 Loss CRIP2

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          hemopoiesis
          zinc ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline