Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03889709_cn
          See other CRIP2 CNV Assays ›
          Gene Symbol
          CRIP2
          Assay Reference Genome
          Location

          Chr.14:105476310 on build GRCh38
          Cytoband
          14q32.33
          Assay ID Hs03889709_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1397

          Gene Name:

          cysteine rich protein 2

          Gene Aliases:

          CRIP, CRP2, ESP1

          Location:

          Chr.14:105472938-105480170 on Build GRCh38

          Assay Gene Location:

          Within Exon 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CRIP2 NM_001270837.1 NP_001257766.1
          NM_001270841.1 NP_001257770.1
          NM_001312.3 NP_001303.1
          NR_073081.1
          NR_073082.1
          NR_073083.1
          NR_073084.1 1 452
          AK057837.1
          AK091845.1 1 452
          AK223251.1 BAD96971.1
          AK300092.1
          AK315757.1
          BC000434.2 AAH00434.1
          BC001931.1 AAH01931.1
          BC034151.1 AAH34151.1
          BC128101.1
          BE271979.1
          BT019911.1 AAV38714.1
          CN367966.1
          D42123.1 BAA07703.1
          HY078632.1
          HY161884.1
          U36190.1 AAB03194.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv3996n54 Chr.14:105474226 - 105488519 on Build GRCh38 Gain CRIP1 CRIP2
          nsv566204 Chr.14:105376842 - 105479048 on Build GRCh38 Gain PACS2 LOC100507437 TEX22 MTA1 CRIP2
          nsv817647 Chr.14:105382554 - 105480505 on Build GRCh38 Gain PACS2 LOC100507437 TEX22 MTA1 CRIP2
          dgv3998n54 Chr.14:105474495 - 105770768 on Build GRCh38 Loss IGHA1 IGHG1 LOC107987211 IGH IGHG3 LOC105370697 C14orf80 ELK2AP MIR8071-1 MIR8071-2 IGHA2 IGHG2 TMEM121 CRIP1 CRIP2 IGHG4
          nsv1160204 Chr.14:105291411 - 105483783 on Build GRCh38 Duplication PACS2 LOC100507437 TEX22 MTA1 CRIP2 BRF1
          dgv3994n54 Chr.14:105474172 - 105477662 on Build GRCh38 Gain CRIP2
          nsv566259 Chr.14:105474444 - 105477342 on Build GRCh38 Gain+Loss CRIP2
          nsv566206 Chr.14:105439736 - 105543235 on Build GRCh38 Loss MTA1 TMEM121 CRIP1 CRIP2 LOC105370697 C14orf80
          nsv832890 Chr.14:105304955 - 105483089 on Build GRCh38 Loss PACS2 LOC100507437 TEX22 MTA1 CRIP2 BRF1
          nsv1440 Chr.14:105473170 - 105479862 on Build GRCh38 Insertion CRIP2
          nsv566210 Chr.14:105465248 - 105488368 on Build GRCh38 Gain MTA1 CRIP1 CRIP2
          nsv827088 Chr.14:105454469 - 105498549 on Build GRCh38 Gain MTA1 CRIP1 CRIP2 C14orf80
          nsv952282 Chr.14:105363764 - 105510163 on Build GRCh38 Deletion PACS2 LOC100507437 TEX22 MTA1 CRIP1 CRIP2 C14orf80
          dgv3999n54 Chr.14:105474867 - 105487998 on Build GRCh38 Loss CRIP1 CRIP2
          esv2760014 Chr.14:105047009 - 105483089 on Build GRCh38 Loss PACS2 GPR132 LOC100507437 LOC102723354 NUDT14 TEX22 MIR6765 MTA1 CRIP2 JAG2 BRF1 BTBD6
          dgv3988n54 Chr.14:105466757 - 105477662 on Build GRCh38 Gain MTA1 CRIP2
          nsv470679 Chr.14:105124547 - 105605978 on Build GRCh38 Loss PACS2 LOC100507437 NUDT14 IGH MIR6765 JAG2 BRF1 LOC105370697 C14orf80 IGHA2 TEX22 MTA1 TMEM121 CRIP1 CRIP2 BTBD6
          dgv3987n54 Chr.14:105461599 - 105499970 on Build GRCh38 Loss MTA1 CRIP1 CRIP2 C14orf80
          nsv566265 Chr.14:105474766 - 105479048 on Build GRCh38 Loss CRIP2
          nsv1054190 Chr.14:105246882 - 105701138 on Build GRCh38 Gain PACS2 LOC100507437 IGH BRF1 LOC105370697 C14orf80 ELK2AP MIR8071-1 MIR8071-2 IGHA2 TEX22 IGHG2 MTA1 TMEM121 CRIP1 CRIP2 BTBD6 IGHG4
          esv3635645 Chr.14:105473682 - 105484540 on Build GRCh38 Gain CRIP2
          nsv517055 Chr.14:105136282 - 106779068 on Build GRCh38 Gain+Loss IGHV3-69-1 PACS2 ADAM6 IGHG1 IGHV4-31 LOC107987211 MIR6765 IGHG3 MIR4537 LOC105370697 IGHV3-71 C14orf80 MIR8071-1 IGHV3-30 IGHD MTA1 IGHV4-55 IGHG4 IGHA1 LOC100507437 IGHV3-41 NUDT14 IGH MIR4538 KIAA0125 JAG2 BRF1 IGHV1-69 LINC00226 LINC00221 MIR4539 ELK2AP MIR8071-2 IGHM IGHA2 TEX22 IGHG2 IGHV3-48 TMEM121 IGHV3-23 CRIP1 CRIP2 BTBD6 MIR4507
          dgv3991n54 Chr.14:105472502 - 105480505 on Build GRCh38 Loss CRIP2

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs78427968] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          hemopoiesis
          zinc ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline