Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03899540_cn
          See other AKAP13 CNV Assays ›
          Gene Symbol
          AKAP13
          Assay Reference Genome
          Location

          Chr.15:85410575 on build GRCh38
          Cytoband
          Assay ID Hs03899540_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          11214

          Gene Name:

          A-kinase anchoring protein 13

          Gene Aliases:

          AKAP-13, AKAP-Lbc, ARHGEF13, BRX, HA-3, Ht31, LBC, PRKA13, PROTO-LB, PROTO-LBC, c-lbc, p47

          Location:

          Chr.15:85380603-85749358 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          AKAP13 NM_006738.6 NP_006729.4
          NM_007200.5 NP_009131.2
          AB055890.1 BAB62913.1
          AF406992.1 AAL11723.1
          AK125331.1
          BC050312.1 AAH50312.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv4692n54 Chr.15:85235254 - 85600273 on Build GRCh38 Gain MIR7706 AKAP13 GOLGA6L3P
          nsv6501969 Chr.15:85407701 - 85410900 on Build GRCh38 Deletion AKAP13
          esv2761699 Chr.15:85236347 - 85569198 on Build GRCh38 Gain MIR7706 AKAP13 GOLGA6L3P
          esv2656709 Chr.15:85406374 - 85411008 on Build GRCh38 Deletion AKAP13
          nsv6514724 Chr.15:85405301 - 85411000 on Build GRCh38 Deletion AKAP13
          dgv2648n100 Chr.15:85268380 - 85605207 on Build GRCh38 Gain MIR7706 AKAP13
          dgv323n27 Chr.15:85282336 - 85600273 on Build GRCh38 Gain MIR7706 AKAP13
          esv3637112 Chr.15:85384885 - 85468079 on Build GRCh38 Gain AKAP13
          esv3637114 Chr.15:85406372 - 85411010 on Build GRCh38 Loss AKAP13
          nsv518064 Chr.15:85178426 - 85605928 on Build GRCh38 Gain MIR7706 LOC101929479 AKAP13 GOLGA6L3P LOC727751
          nsv4531528 Chr.15:85213768 - 85599769 on Build GRCh38 Duplication MIR7706 LOC101929479 AKAP13 GOLGA6L3P LOC727751
          nsv5527698 Chr.15:85406376 - 85411008 on Build GRCh38 Deletion AKAP13
          dgv4694n54 Chr.15:85344022 - 85600273 on Build GRCh38 Gain MIR7706 AKAP13
          nsv570298 Chr.15:85272357 - 85510128 on Build GRCh38 Gain MIR7706 AKAP13
          esv3637111 Chr.15:85291627 - 85464874 on Build GRCh38 Gain MIR7706 AKAP13
          esv21779 Chr.15:85406498 - 85411061 on Build GRCh38 Loss AKAP13
          dgv754e212 Chr.15:85181918 - 85597222 on Build GRCh38 Gain MIR7706 LOC101929479 AKAP13 GOLGA6L3P LOC727751
          nsv428311 Chr.15:85107707 - 85486274 on Build GRCh38 Gain PDE8A MIR7706 LOC101929479 AKAP13 GOLGA6L3P LOC727751

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          G-protein coupled receptor signaling pathway
          heart development
          regulation of Rho protein signal transduction
          positive regulation of Rho protein signal transduction
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of small GTPase mediated signal transduction
          cardiac muscle cell differentiation
          regulation of sarcomere organization
          bone development
          adrenergic receptor signaling pathway
          regulation of intracellular signal transduction
          MAP-kinase scaffold activity
          guanyl-nucleotide exchange factor activity
          protein binding
          zinc ion binding
          small GTPase binding
          metal ion binding
          protein kinase A binding
          binding, bridging

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline