Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03910383_cn
          See other SORD CNV Assays ›
          Gene Symbol
          SORD
          Assay Reference Genome
          Location

          Chr.15:45025174 on build GRCh38
          Cytoband
          Assay ID Hs03910383_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          6652

          Gene Name:

          sorbitol dehydrogenase

          Gene Aliases:

          HEL-S-95n, HMNR8, RDH, SDH, SORD1, SORDD, XDH

          Location:

          Chr.15:45023195-45077185 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SORD NM_003104.6 NP_003095.2
          NR_034039.2
          AK295656.1 BAH12142.1
          AK296574.1 BAG59194.1
          AK309672.1
          AK312444.1 BAG35352.1
          BC021085.2 AAH21085.1
          BC025295.1 AAH25295.1
          DB031906.1
          FJ224316.1 ACI46008.1
          L29008.1 AAA80565.1
          U07361.1 AAA66064.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4241837 Chr.15:45024150 - 45027138 on Build GRCh38 Deletion SORD
          dgv2590n100 Chr.15:44860173 - 45034007 on Build GRCh38 Gain SORD SORD2P TERB2
          nsv977710 Chr.15:45023993 - 45068797 on Build GRCh38 Duplication SORD
          nsv509570 Chr.15:44997984 - 45057142 on Build GRCh38 Insertion SORD
          esv2760029 Chr.15:44676670 - 45130644 on Build GRCh38 Gain+Loss DUOXA2 B2M DUOX1 TRIM69 PATL2 SORD DUOXA1 RNF4BP MIR10393 SORD2P DUOX2 TERB2
          nsv4748065 Chr.15:44857549 - 45026631 on Build GRCh38 Deletion SORD SORD2P TERB2
          nsv9259 Chr.15:45025170 - 45034152 on Build GRCh38 Gain SORD

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose metabolic process
          sorbitol metabolic process
          sorbitol catabolic process
          glucuronate catabolic process to xylulose 5-phosphate
          sperm motility
          fructose biosynthetic process
          L-xylitol catabolic process
          L-xylitol metabolic process
          (R,R)-butanediol dehydrogenase activity
          L-iditol 2-dehydrogenase activity
          alcohol dehydrogenase (NAD) activity
          zinc ion binding
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          carbohydrate binding
          identical protein binding
          D-xylulose reductase activity
          metal ion binding
          D-sorbitol dehydrogenase (acceptor) activity
          ribitol 2-dehydrogenase activity
          NAD binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline