Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03931538_cn
          See other TCF25 CNV Assays ›
          Gene Symbol
          TCF25
          Assay Reference Genome
          Location

          Chr.16:89876181 on build GRCh38
          Cytoband
          Assay ID Hs03931538_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          22980

          Gene Name:

          TCF25 ribosome quality control complex subunit

          Gene Aliases:

          FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049

          Location:

          Chr.16:89873592-89911379 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TCF25 NM_014972.3 NP_055787.1
          AF322111.1 AAG50276.1
          AK024679.1 BAB14959.1
          AK301053.1 BAG62664.1
          BC000959.1 AAH00959.1
          BC009349.2 AAH09349.1
          BC011884.2 AAH11884.1
          BI913551.1
          DQ321703.1 ABC55265.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv457624 Chr.16:89738447 - 89915950 on Build GRCh38 Loss FANCA ZNF276 LOC124903759 SPIRE2 TCF25
          esv2762221 Chr.16:89610380 - 90039318 on Build GRCh38 Gain+Loss FANCA ZNF276 SNORA119 VPS9D1 CHMP1A DEF8 SPATA33 DBNDD1 CENPBD1P GAS8-AS1 AFG3L1P CDK10 DPEP1 LINC02166 VPS9D1-AS1 SPATA2L LOC124903759 TUBB3 SPIRE2 DRC4 MC1R TCF25
          esv2758671 Chr.16:89804324 - 89901541 on Build GRCh38 Loss FANCA SPIRE2 TCF25
          nsv5928506 Chr.16:89860057 - 89893275 on Build GRCh38 Duplication SPIRE2 TCF25
          nsv5938953 Chr.16:89875732 - 89878878 on Build GRCh38 Deletion TCF25
          nsv4245632 Chr.16:89844192 - 89887592 on Build GRCh38 Deletion SPIRE2 TCF25
          nsv573810 Chr.16:89857498 - 90037251 on Build GRCh38 Loss GAS8-AS1 AFG3L1P SNORA119 DEF8 LOC124903759 TUBB3 DBNDD1 SPIRE2 DRC4 MC1R CENPBD1P TCF25
          nsv428331 Chr.16:89216635 - 90185980 on Build GRCh38 Gain+Loss SPG7 ZNF276 CPNE7 VPS9D1 ZNF778 CHMP1A DEF8 PRDM7 TUBB8P7 DBNDD1 LOC128462377 CENPBD1P CDK10 DPEP1 LOC100287036 LINC02166 VPS9D1-AS1 FAM157C LOC105376781 SPATA2L LOC124903757 TUBB3 SNORD68 MC1R FANCA SNORA119 LOC101927817 SPATA33 RPL13 GAS8-AS1 AFG3L1P LOC124903766 LOC105371414 ZNF778-DT LOC101927863 ANKRD11 URAHP LOC124903759 SPIRE2 DRC4 TCF25
          nsv6500661 Chr.16:89839303 - 90017373 on Build GRCh38 Deletion AFG3L1P SNORA119 DEF8 LOC124903759 TUBB3 DBNDD1 SPIRE2 MC1R CENPBD1P TCF25
          nsv573799 Chr.16:89852186 - 90022379 on Build GRCh38 Gain AFG3L1P SNORA119 DEF8 LOC124903759 TUBB3 DBNDD1 SPIRE2 DRC4 MC1R CENPBD1P TCF25
          nsv482951 Chr.16:88633593 - 90228345 on Build GRCh38 Loss ZNF276 LINC00304 CBFA2T3 VPS9D1 ZNF778 CHMP1A DEF8 TUBB8P7 DBNDD1 SNAI3 TRAPPC2L LINC02193 LOC100287036 CYBA VPS9D1-AS1 SPATA2L TUBB3 SNORD68 SNAI3-AS1 PABPN1L MVD SNORA119 LOC101927817 SPATA33 RPL13 HSALR1 GAS8-AS1 CDH15 LOC101927863 ANKRD11 URAHP SPIRE2 DRC4 SLC22A31 TCF25 LINC02138 SPG7 CPNE7 ACSF3 PRDM7 LOC128462377 GALNS CENPBD1P CDK10 DPEP1 RNF166 LINC02166 FAM157C LOC105376781 LOC124903757 MC1R FANCA LOC100129697 PIEZO1-AS1 APRT PIEZO1 LOC101927793 AFG3L1P CDT1 LOC124903766 LOC105371414 ZNF778-DT CTU2 LOC124903759 MIR4722 IL17C

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein K48-linked ubiquitination
          rescue of stalled ribosome
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline