Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03936147_cn
          See other TCF25 CNV Assays ›
          Gene Symbol
          TCF25
          Assay Reference Genome
          Location

          Chr.16:89897461 on build GRCh38
          Cytoband
          Assay ID Hs03936147_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          22980

          Gene Name:

          TCF25 ribosome quality control complex subunit

          Gene Aliases:

          FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049

          Location:

          Chr.16:89873592-89911379 on Build GRCh38

          Assay Gene Location:

          Within Intron 9
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TCF25 NM_014972.3 NP_055787.1
          AB028972.1 BAA83001.1
          AF130092.1 AAG35518.1
          AF322111.1 AAG50276.1
          BC000959.1 AAH00959.1
          BC009349.2 AAH09349.1
          BC011884.2 AAH11884.1
          DQ321703.1 ABC55265.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv457624 Chr.16:89738447 - 89915950 on Build GRCh38 Loss LOC124903759 ZNF276 FANCA SPIRE2 TCF25
          esv2762221 Chr.16:89610380 - 90039318 on Build GRCh38 Gain+Loss MC1R DBNDD1 LOC124903759 ZNF276 GAS8-AS1 VPS9D1-AS1 SPATA2L SPIRE2 CHMP1A CENPBD1P DRC4 AFG3L1P TUBB3 DEF8 FANCA CDK10 VPS9D1 SPATA33 LINC02166 TCF25 DPEP1 SNORA119
          esv2758671 Chr.16:89804324 - 89901541 on Build GRCh38 Loss FANCA SPIRE2 TCF25
          dgv3359n152 Chr.16:89883326 - 89919832 on Build GRCh38 Insertion MC1R LOC124903759 TCF25
          nsv573810 Chr.16:89857498 - 90037251 on Build GRCh38 Loss TUBB3 MC1R DBNDD1 LOC124903759 DEF8 GAS8-AS1 SPIRE2 CENPBD1P TCF25 DRC4 AFG3L1P SNORA119
          nsv428331 Chr.16:89216635 - 90185980 on Build GRCh38 Gain+Loss DBNDD1 LOC124903759 PRDM7 GAS8-AS1 TUBB8P7 LOC128462377 LOC100287036 DRC4 AFG3L1P SPG7 LOC124903757 SNORD68 DEF8 LOC105376781 ANKRD11 FANCA VPS9D1 LINC02166 TCF25 RPL13 DPEP1 SNORA119 MC1R ZNF276 LOC101927863 VPS9D1-AS1 ZNF778-DT LOC105371414 SPATA2L SPIRE2 CHMP1A CENPBD1P ZNF778 TUBB3 FAM157C LOC101927817 LOC124903766 URAHP CDK10 CPNE7 SPATA33
          nsv6500661 Chr.16:89839303 - 90017373 on Build GRCh38 Deletion TUBB3 MC1R DBNDD1 LOC124903759 DEF8 SPIRE2 CENPBD1P TCF25 AFG3L1P SNORA119
          nsv573799 Chr.16:89852186 - 90022379 on Build GRCh38 Gain TUBB3 MC1R DBNDD1 LOC124903759 DEF8 SPIRE2 CENPBD1P TCF25 DRC4 AFG3L1P SNORA119
          nsv482951 Chr.16:88633593 - 90228345 on Build GRCh38 Loss DBNDD1 LOC124903759 PRDM7 TUBB8P7 LOC128462377 APRT AFG3L1P GALNS SNORD68 DEF8 LOC105376781 VPS9D1 RPL13 DPEP1 LOC101927863 SPIRE2 CHMP1A LOC101927793 CDH15 MVD LOC101927817 HSALR1 CPNE7 SPATA33 PIEZO1 MIR4722 LINC02193 IL17C LINC02138 GAS8-AS1 LOC100287036 LINC00304 DRC4 SPG7 LOC124903757 RNF166 ANKRD11 FANCA LOC100129697 LINC02166 TCF25 SNORA119 MC1R ZNF276 VPS9D1-AS1 SLC22A31 TRAPPC2L ZNF778-DT LOC105371414 CDT1 SPATA2L PIEZO1-AS1 CENPBD1P ZNF778 SNAI3 PABPN1L TUBB3 CBFA2T3 FAM157C ACSF3 SNAI3-AS1 LOC124903766 CYBA URAHP CDK10 CTU2
          nsv1066499 Chr.16:89891957 - 90028891 on Build GRCh38 Loss TUBB3 MC1R DBNDD1 LOC124903759 DEF8 CENPBD1P TCF25 DRC4 AFG3L1P SNORA119

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein K48-linked ubiquitination
          rescue of stalled ribosome
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline