Search
Search

Entrez Gene ID: | 8714 |
Gene Name: | ATP binding cassette subfamily C member 3 |
Gene Aliases: |
ABC31, EST90757, MLP2, MOAT-D, MRP3, cMOAT2 |
Location: |
Chr.17:50634843-50691704 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCC3 | NM_003786.3 | NP_003777.2 | ||
| XM_005257763.3 | XP_005257820.1 | |||
| XM_011525422.2 | XP_011523724.1 | |||
| XM_011525423.1 | XP_011523725.1 | |||
| XM_011525424.2 | XP_011523726.1 | |||
| XM_011525425.2 | XP_011523727.1 | |||
| XM_017025265.1 | XP_016880754.1 | |||
| XM_017025266.1 | XP_016880755.1 | |||
| AB010887.1 | BAA28146.1 | |||
| AB208954.1 | BAD92191.1 | |||
| AF009670.1 | AAD01430.1 | |||
| AF083552.1 | AAC34668.1 | |||
| AF085690.1 | AAD02845.1 | |||
| AF085691.1 | AAD02846.1 | |||
| AF085692.1 | AAD02847.1 | |||
| AF104943.1 | AAD04170.1 | |||
| BC137347.1 | ||||
| BC137348.1 | ||||
| Y17151.2 | CAA76658.2 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv42n68 | Chr.17:50584511 - 50823099 on Build GRCh38 | Loss |
|
| nsv833477 | Chr.17:50516880 - 50678469 on Build GRCh38 | Loss |
|
Set Membership: |
|