Search
Search

Entrez Gene ID: | 284058 |
Gene Name: | KAT8 regulatory NSL complex subunit 1 |
Gene Aliases: |
C17DELq21.31, CENP-36, DEL17Q21.31, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1 |
Location: |
Chr.17:732028-927537 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5619n54 | Chr.17:46040903 - 46215654 on Build GRCh38 | Gain+Loss |
|
| nsv4253669 | Chr.17:45994634 - 46073634 on Build GRCh38 | Duplication |
|
| nsv1072740 | Chr.17:45970433 - 46248034 on Build GRCh38 | Deletion |
|
| esv2751686 | Chr.17:45962198 - 46286690 on Build GRCh38 | Gain |
|
| esv2758692 | Chr.17:46006548 - 47199967 on Build GRCh38 | Gain+Loss |
|
| nsv1057963 | Chr.17:46039494 - 46215377 on Build GRCh38 | Gain |
|
| dgv128e55 | Chr.17:46017105 - 46286690 on Build GRCh38 | Gain |
|
| nsv469705 | Chr.17:45969304 - 46127121 on Build GRCh38 | Loss |
|
| nsv471698 | Chr.17:46002113 - 46300887 on Build GRCh38 | Gain+Loss |
|
| nsv469537 | Chr.17:46002112 - 46163353 on Build GRCh38 | Gain+Loss |
|
| nsv575142 | Chr.17:46003698 - 46172742 on Build GRCh38 | Loss |
|
| nsv2069 | Chr.17:46034385 - 46065659 on Build GRCh38 | Insertion |
|
| esv2762433 | Chr.17:45496768 - 46710828 on Build GRCh38 | Gain+Loss |
|
| dgv366n27 | Chr.17:46032477 - 46215654 on Build GRCh38 | Gain |
|
| dgv5618n54 | Chr.17:46032108 - 46330700 on Build GRCh38 | Gain |
|
| dgv127e55 | Chr.17:45927606 - 46733750 on Build GRCh38 | Gain |
|
Set Membership: |
|