Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03968388_cn
          See other AATK CNV Assays ›
          Gene Symbol
          AATK
          Assay Reference Genome
          Location

          Chr.17:81135652 on build GRCh38
          Cytoband
          17q25.3
          Assay ID Hs03968388_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          9625

          Gene Name:

          apoptosis associated tyrosine kinase

          Gene Aliases:

          AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP

          Location:

          Chr.17:81117295-81166072 on Build GRCh38

          Assay Gene Location:

          Within Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          AATK NM_001080395.2 NP_001073864.2
          XM_006722193.2 XP_006722256.1
          XM_006722194.2 XP_006722257.1
          XM_006722196.2 XP_006722259.1
          XM_017025421.1 XP_016880910.1
          AK131529.1 BAD18667.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv26223 Chr.17:80659041 - 81736487 on Build GRCh38 Gain+Loss MIR3065 MIR1250 BAIAP2-AS1 AATK-AS1 MIR3186 MRPL12 LOC100130370 MIR4740 TMEM105 ARL16 FSCN2 LOC105371925 C17orf89 HGS LOC101928855 MIR6786 CEP131 SLC25A10 LOC100129503 MIR657 LINC00482 BAIAP2 MIR338 CHMP6 ACTG1 AATK FAAP100 BAHCC1 OXLD1 LOC107985021 NPLOC4 TEPSIN SLC38A10 LOC105371921 TSPAN10 LOC400627 RPTOR PDE6G LOC107985022 CCDC137
          dgv46n68 Chr.17:81133921 - 81288932 on Build GRCh38 Loss TEPSIN SLC38A10 AATK LOC105371925 C17orf89 AATK-AS1 CEP131
          esv3641392 Chr.17:81126529 - 81136632 on Build GRCh38 Gain AATK MIR1250
          nsv1065052 Chr.17:81080876 - 81204920 on Build GRCh38 Gain AATK MIR3065 MIR1250 AATK-AS1 CEP131 MIR657 BAIAP2 MIR338
          nsv833564 Chr.17:81014713 - 81164054 on Build GRCh38 Loss AATK MIR3065 MIR1250 BAIAP2-AS1 MIR657 BAIAP2 MIR338
          nsv470619 Chr.17:81032233 - 81349843 on Build GRCh38 Loss AATK MIR3065 MIR1250 BAIAP2-AS1 AATK-AS1 TEPSIN TMEM105 SLC38A10 LOC105371925 C17orf89 CEP131 MIR657 LINC00482 BAIAP2 MIR338
          dgv3292n100 Chr.17:81019582 - 81204920 on Build GRCh38 Gain AATK MIR3065 MIR1250 BAIAP2-AS1 AATK-AS1 CEP131 MIR657 BAIAP2 MIR338
          nsv1066947 Chr.17:81094837 - 81181290 on Build GRCh38 Loss AATK MIR3065 MIR1250 AATK-AS1 MIR657 BAIAP2 MIR338
          nsv523417 Chr.17:81115790 - 81164271 on Build GRCh38 Gain AATK MIR3065 MIR1250 MIR657 BAIAP2 MIR338
          nsv952388 Chr.17:81034701 - 81137300 on Build GRCh38 Deletion AATK MIR3065 MIR1250 BAIAP2-AS1 MIR657 BAIAP2 MIR338

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs78064897] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          protein serine/threonine kinase activity
          ATP binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline