Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03999235_cn
          See other SERPINB2 CNV Assays ›
          Gene Symbol
          SERPINB2
          Assay Reference Genome
          Location

          Chr.18:63902441 on build GRCh38
          Cytoband
          Assay ID Hs03999235_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5055

          Gene Name:

          serpin family B member 2

          Gene Aliases:

          HsT1201, PAI, PAI-2, PAI2, PLANH2

          Location:

          Chr.18:63887705-63903888 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 6 - Exon 7
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SERPINB2 NM_001143818.2 NP_001137290.1
          NM_002575.3 NP_002566.1
          XM_024451192.2 XP_024306960.1
          A21238.1 CAA01535.1
          A32415.1 CAA02099.1
          AK313159.1 BAG35977.1
          BC012609.1 AAH12609.1
          J02685.1 AAA36413.1
          J03603.1 AAA60004.1
          M18082.1 AAA60006.1
          Y00630.1 CAA68666.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1066552 Chr.18:63147299 - 64608176 on Build GRCh38 Gain SERPINB7 SERPINB5 SERPINB8 SERPINB13 SERPINB11 BCL2 LINC01924 SERPINB12 SERPINB2 KDSR-DT KDSR SERPINB4 LINC00305 LINC01538 SERPINB3 SERPINB10 VPS4B HMSD
          nsv6526003 Chr.18:63713430 - 64357552 on Build GRCh38 Deletion SERPINB2 SERPINB7 SERPINB8 SERPINB11 LINC00305 LINC01538 LINC01924 SERPINB10 HMSD
          nsv5514480 Chr.18:63714707 - 64278418 on Build GRCh38 Duplication SERPINB2 SERPINB7 SERPINB8 SERPINB11 LINC00305 LINC01538 LINC01924 SERPINB10 HMSD
          nsv833673 Chr.18:63897072 - 64097021 on Build GRCh38 Gain SERPINB2 SERPINB8 LINC00305 SERPINB10 HMSD
          nsv4532319 Chr.18:63817764 - 66099664 on Build GRCh38 Deletion SERPINB2 CDH7 SERPINB8 LINC00305 LINC01538 LINC01924 SERPINB10 HMSD
          nsv6519660 Chr.18:63146370 - 64608618 on Build GRCh38 Duplication SERPINB7 SERPINB5 SERPINB8 SERPINB13 SERPINB11 BCL2 LINC01924 SERPINB12 SERPINB2 KDSR-DT KDSR SERPINB4 LINC00305 LINC01538 SERPINB3 SERPINB10 VPS4B HMSD
          esv21739 Chr.18:63715122 - 64278129 on Build GRCh38 Gain SERPINB2 SERPINB7 SERPINB8 SERPINB11 LINC00305 LINC01538 LINC01924 SERPINB10 HMSD
          esv3642785 Chr.18:63831255 - 63986077 on Build GRCh38 Gain SERPINB2 SERPINB8 SERPINB10 HMSD
          nsv1067311 Chr.18:63818011 - 66637685 on Build GRCh38 Loss SERPINB2 CDH7 SERPINB8 LINC00305 LINC01538 LINC01924 CDH19 SERPINB10 HMSD
          nsv6520851 Chr.18:63775316 - 64080261 on Build GRCh38 Duplication SERPINB2 SERPINB7 SERPINB8 LINC00305 SERPINB10 HMSD
          nsv525889 Chr.18:63845132 - 63948110 on Build GRCh38 Loss SERPINB2 SERPINB10
          esv2758730 Chr.18:63881807 - 63923851 on Build GRCh38 Loss SERPINB2 SERPINB10

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          fibrinolysis
          negative regulation of apoptotic process
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline