Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04008514_cn
          See other PEX11G CNV Assays ›
          Gene Symbol
          PEX11G
          Assay Reference Genome
          Location

          Chr.19:7480325 on build GRCh38
          Cytoband
          Assay ID Hs04008514_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          92960

          Gene Name:

          peroxisomal biogenesis factor 11 gamma

          Gene Aliases:

          PEX11gamma

          Location:

          Chr.19:7476875-7494977 on Build GRCh38

          Assay Gene Location:

          Within Intron 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PEX11G NM_001270539.2 NP_001257468.1
          NM_001300881.2 NP_001287810.1
          NM_080662.4 NP_542393.1
          XM_005272506.3 XP_005272563.1
          XM_011528426.3 XP_011526728.1
          XM_011528427.3 XP_011526729.1
          XM_011528428.1 XP_011526730.1
          XM_011528429.3 XP_011526731.1
          XM_011528431.3 XP_011526733.1
          XM_011528432.2 XP_011526734.1
          AB095921.1 BAD01558.1
          AL833945.1 CAD38800.1
          BC008780.2 AAH08780.1
          BI833221.1
          HY002496.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1060887 Chr.19:7089546 - 7861087 on Build GRCh38 Gain CLEC4G FCER2 CAMSAP3 MCEMP1 TRAPPC5 CLEC4GP1 ZNF358 SAXO5 PEX11G XAB2 ARHGEF18-AS1 ARHGEF18 PCP2 INSR MIR6792 MCOLN1 STXBP2 CD209 EVI5L PET100 CLEC4M PNPLA6 RETN
          nsv509719 Chr.19:7390693 - 7484911 on Build GRCh38 Insertion PEX11G ARHGEF18-AS1 ARHGEF18
          esv2751809 Chr.19:6896369 - 7816144 on Build GRCh38 Gain MARK2P21 MBD3L3 SAXO5 PEX11G ARHGEF18-AS1 PCP2 ADGRE1 INSR MIR6792 MCOLN1 STXBP2 CD209 PET100 RETN MBD3L2B CLEC4G MBD3L5 ZNF557 FCER2 CAMSAP3 MCEMP1 TRAPPC5 ADGRE4P CLEC4GP1 ZNF358 XAB2 ARHGEF18 MBD3L4 MBD3L2 CLEC4M PNPLA6
          nsv4261533 Chr.19:7476696 - 7481020 on Build GRCh38 Deletion PEX11G ARHGEF18

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs111618462,rs73488445] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          peroxisome fission
          regulation of peroxisome size
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline