Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04026631_cn
          See other CACNG8 CNV Assays ›
          Gene Symbol
          CACNG8
          Assay Reference Genome
          Location

          Chr.19:53984044 on build GRCh38
          Cytoband
          19q13.42
          Assay ID Hs04026631_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          59283

          Gene Name:

          calcium voltage-gated channel auxiliary subunit gamma 8

          Gene Aliases:

          -

          Location:

          Chr.19:53963036-53990215 on Build GRCh38

          Assay Gene Location:

          Within Exon 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CACNG8 NM_031895.5 4 2580 NP_114101.4

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv458782 Chr.19:53961144 - 54093354 on Build GRCh38 Gain CACNG6 VSTM1 MIR935 TARM1 CACNG8
          nsv1063127 Chr.19:53926876 - 54068977 on Build GRCh38 Gain CACNG6 VSTM1 MIR935 CACNG7 CACNG8
          esv2718812 Chr.19:53184636 - 54322450 on Build GRCh38 Deletion MIR519B MIR527 MIR520E TFPT LOC284379 ZNF818P MIR523 VN1R2 MIR373 ZNF845 TMC4 PRKCG MIR518A2 MIR515-2 MIR518B MIR1283-1 MIR520H LOC105372457 NDUFA3 MIR516B2 CACNG6 MIR518D MIR519E MIR520A OSCAR ZNF525 CACNG7 ZNF761 LILRB5 MIR526B MIR371B RPS9 LILRB2 MIR518E ZNF813 MIR524 VN1R4 MIR519D MIR1283-2 MIR521-1 MIR515-1 MIR520B TSEN34 DPRX ZNF677 TARM1 LENG1 LILRA6 ZNF331 MIR525 VSTM1 MIR522 MIR517B MIR526A1 MIR521-2 MIR520G MIR516B1 MIR526A2 MIR518A1 MIR518F MIR517C MIR935 MIR518C BIRC8 MIR520D CACNG8 MIR520F ZNF665 MIR519A2 LILRA5 CNOT3 NLRP12 MBOAT7 MYADM MIR517A TPM3P9 PRPF31 MIR372 MIR512-2 MIR520C MIR512-1 MIR498 LILRB3 MIR4752 MIR371A MIR519A1 ZNF765 MIR519C MIR516A1 MIR516A2 FAM90A27P MIR1323
          nsv580111 Chr.19:53977896 - 53989668 on Build GRCh38 Loss MIR935 CACNG8
          nsv953612 Chr.19:53978147 - 53994046 on Build GRCh38 Deletion CACNG6 MIR935 CACNG8
          nsv833876 Chr.19:53845956 - 53995117 on Build GRCh38 Loss CACNG6 MIR935 PRKCG CACNG7 MYADM CACNG8 LOC105372457
          nsv833877 Chr.19:53951529 - 54117323 on Build GRCh38 Loss CACNG6 VSTM1 MIR935 OSCAR TARM1 TFPT CACNG8 PRPF31 NDUFA3

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          transmission of nerve impulse
          cardiac conduction
          calcium ion transmembrane transport
          regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          voltage-gated calcium channel activity
          channel regulator activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline