Search
Search

Entrez Gene ID: | 9215 |
Gene Name: | LARGE xylosyl- and glucuronyltransferase 1 |
Gene Aliases: |
LARGE, MDC1D, MDDGA6, MDDGB6 |
Location: |
Chr.22:33066663-33922824 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055791 | Chr.22:33426240 - 33763549 on Build GRCh38 | Loss |
|
| nsv588932 | Chr.22:33470868 - 33536638 on Build GRCh38 | Gain |
|
| nsv1064026 | Chr.22:33371199 - 33506073 on Build GRCh38 | Loss |
|
| nsv6538829 | Chr.22:33421845 - 33559203 on Build GRCh38 | Deletion |
|
| nsv6543068 | Chr.22:33501650 - 33525201 on Build GRCh38 | Deletion |
|
| nsv6546779 | Chr.22:33359964 - 33503028 on Build GRCh38 | Deletion |
|
| esv2758832 | Chr.22:33472959 - 33513560 on Build GRCh38 | Gain+Loss |
|
| nsv6552934 | Chr.22:33473880 - 33680313 on Build GRCh38 | Deletion |
|
| esv2763261 | Chr.22:33435362 - 33892401 on Build GRCh38 | Loss |
|
| esv3568379 | Chr.22:33458829 - 33815605 on Build GRCh38 | Loss |
|
| dgv4562n100 | Chr.22:33451090 - 33538478 on Build GRCh38 | Gain |
|
| esv2751963 | Chr.22:33459526 - 33539911 on Build GRCh38 | Gain |
|
| nsv6537656 | Chr.22:33466211 - 33538664 on Build GRCh38 | Duplication |
|
| nsv5544834 | Chr.22:33493300 - 33504421 on Build GRCh38 | Deletion |
|
| nsv588931 | Chr.22:33438682 - 33511978 on Build GRCh38 | Loss |
|
| nsv6546707 | Chr.22:33502301 - 33504300 on Build GRCh38 | Deletion |
|
Set Membership: |
|