Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04211858_cn
          See other FCGR3B CNV Assays ›
          Gene Symbol
          FCGR3B
          Assay Reference Genome
          Location

          Chr.1:161626930 on build GRCh38
          Cytoband
          1q23.3
          Assay ID Hs04211858_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          2215

          Gene Name:

          Fc fragment of IgG receptor IIIb

          Gene Aliases:

          CD16, CD16b, FCG3, FCGR3, FCR-10, FCRIII, FCRIIIb

          Location:

          Chr.1:161623196-161631963 on Build GRCh38

          Assay Gene Location:

          Within Intron 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          FCGR3B NM_000570.4 NP_000561.3
          NM_001244753.1 NP_001231682.1
          NM_001271035.1 NP_001257964.1
          NM_001271036.1 NP_001257965.1
          NM_001271037.1 NP_001257966.1
          AB025256.1 BAA83803.1
          AJ581669.1 CAE46408.1
          AK313219.1
          AK316565.1
          BC128562.1
          DA669913.1
          DA670720.1
          DA671001.1
          DA672751.1
          DA673839.1
          J04162.1 AAA35881.1
          M24854.1 AAA53507.1
          X07934.1 CAA30758.1
          X16863.1 CAA34753.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv819523 Chr.1:161521768 - 161662864 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR2A FCGR3A HSPA6
          esv2507888 Chr.1:161548423 - 161631182 on Build GRCh38 Deletion FCGR2C FCGR3B HSPA7 FCGR3A
          esv3325 Chr.1:161594843 - 161676929 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7
          esv5752 Chr.1:161594815 - 161676980 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7
          nsv517640 Chr.1:161525250 - 161692694 on Build GRCh38 Gain+Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR3A HSPA6 RPL31P11
          esv990743 Chr.1:161550050 - 161649676 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR3A
          esv2758972 Chr.1:161374465 - 161711762 on Build GRCh38 Gain+Loss FCGR2C FCGR2B FCGR3B FCRLA HSPA7 FCGR2A FCGR3A HSPA6 RPL31P11
          esv3587733 Chr.1:161600533 - 161628390 on Build GRCh38 Gain FCGR2C FCGR3B HSPA7
          nsv818500 Chr.1:161541303 - 161647726 on Build GRCh38 Gain FCGR2C FCGR2B FCGR3B HSPA7 FCGR3A
          dgv24n16 Chr.1:161571696 - 161671744 on Build GRCh38 Insertion FCGR2C FCGR2B FCGR3B HSPA7
          nsv548066 Chr.1:161587939 - 161647726 on Build GRCh38 Gain FCGR2C FCGR2B FCGR3B HSPA7
          esv3584532 Chr.1:161541620 - 161669769 on Build GRCh38 Gain FCGR2C FCGR2B FCGR3B HSPA7 FCGR3A
          nsv548074 Chr.1:161623299 - 161671187 on Build GRCh38 Loss FCGR2B FCGR3B
          dgv605n54 Chr.1:161533359 - 161671187 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR3A
          esv28122 Chr.1:161439273 - 161676968 on Build GRCh38 Gain+Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR2A FCGR3A HSPA6
          esv2670711 Chr.1:161620574 - 161628209 on Build GRCh38 Deletion FCGR3B
          nsv428257 Chr.1:161436525 - 161898473 on Build GRCh38 Loss FCGR2B HSPA7 FCRLB ATF6 DUSP12 LOC100422526 FCGR2C FCGR3B FCRLA FCGR2A FCGR3A HSPA6 RPL31P11
          nsv1161862 Chr.1:161539085 - 161678447 on Build GRCh38 Duplication FCGR2C FCGR2B FCGR3B HSPA7 FCGR3A
          dgv606n54 Chr.1:161572474 - 161671187 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7
          nsv820341 Chr.1:161510055 - 161678358 on Build GRCh38 Duplication FCGR2C FCGR2B FCGR3B HSPA7 FCGR2A FCGR3A HSPA6
          dgv447n100 Chr.1:161506699 - 161700539 on Build GRCh38 Gain FCGR2C FCGR2B FCGR3B HSPA7 FCGR2A FCGR3A HSPA6 RPL31P11
          dgv38e201 Chr.1:161558346 - 161653012 on Build GRCh38 Deletion FCGR2C FCGR2B FCGR3B HSPA7
          nsv946468 Chr.1:161594892 - 161670153 on Build GRCh38 Duplication FCGR2C FCGR2B FCGR3B HSPA7
          nsv825254 Chr.1:161607568 - 161649676 on Build GRCh38 Gain FCGR2B FCGR3B HSPA7
          dgv448n100 Chr.1:161527110 - 161668711 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR3A
          esv3541689 Chr.1:161579211 - 161675381 on Build GRCh38 Deletion FCGR2C FCGR2B FCGR3B HSPA7
          esv2718873 Chr.1:161512733 - 161631124 on Build GRCh38 Deletion FCGR2C FCGR3B HSPA7 FCGR2A FCGR3A HSPA6
          esv3587734 Chr.1:161620485 - 161628311 on Build GRCh38 Loss FCGR3B
          dgv23n16 Chr.1:161543533 - 161631305 on Build GRCh38 Insertion FCGR2C FCGR3B HSPA7 FCGR3A
          dgv4n68 Chr.1:161433240 - 161696942 on Build GRCh38 Gain+Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR2A FCGR3A HSPA6 RPL31P11
          nsv464173 Chr.1:161509955 - 161677017 on Build GRCh38 Gain FCGR2C FCGR2B FCGR3B HSPA7 FCGR2A FCGR3A HSPA6
          nsv8502 Chr.1:161507558 - 161676760 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR2A FCGR3A HSPA6
          nsv821621 Chr.1:161553439 - 161667052 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7
          nsv509546 Chr.1:161543042 - 161684642 on Build GRCh38 Insertion FCGR2C FCGR2B FCGR3B HSPA7 FCGR3A RPL31P11
          dgv451n100 Chr.1:161532055 - 161678819 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR3A
          esv997087 Chr.1:161513165 - 161676614 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR2A FCGR3A HSPA6
          dgv85n67 Chr.1:161519583 - 161678358 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR2A FCGR3A HSPA6
          esv33393 Chr.1:161508891 - 162110537 on Build GRCh38 Gain+Loss FCGR2B HSPA7 OLFML2B NOS1AP FCRLB ATF6 DUSP12 LOC100422526 FCGR2C FCGR3B FCRLA FCGR2A FCGR3A HSPA6 RPL31P11
          nsv514915 Chr.1:161539698 - 161676786 on Build GRCh38 Gain FCGR2C FCGR2B FCGR3B HSPA7 FCGR3A
          nsv825242 Chr.1:161589306 - 161649676 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7
          nsv1009190 Chr.1:161623061 - 161649256 on Build GRCh38 Gain+Loss FCGR2B FCGR3B
          dgv5e203 Chr.1:161508746 - 161668740 on Build GRCh38 Gain+Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR2A FCGR3A HSPA6
          dgv75e199 Chr.1:161561480 - 161661159 on Build GRCh38 Deletion FCGR2C FCGR2B FCGR3B HSPA7
          dgv8e55 Chr.1:161493708 - 161703892 on Build GRCh38 Gain FCGR2C FCGR2B FCGR3B HSPA7 FCGR2A FCGR3A HSPA6 RPL31P11
          esv3890693 Chr.1:161521106 - 161654964 on Build GRCh38 Loss FCGR2C FCGR2B FCGR3B HSPA7 FCGR2A FCGR3A HSPA6

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          immune response
          mitophagy in response to mitochondrial depolarization
          xenophagy
          IgG binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0