Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04299567_cn
          See other MAPK9 CNV Assays ›
          Gene Symbol
          MAPK9
          Assay Reference Genome
          Location

          Chr.5:180289162 on build GRCh38
          Cytoband
          5q35.3
          Assay ID Hs04299567_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5601

          Gene Name:

          mitogen-activated protein kinase 9

          Gene Aliases:

          JNK-55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, PRKM9, SAPK, SAPK1a, p54a, p54aSAPK

          Location:

          Chr.5:180233594-180292071 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MAPK9 NM_001135044.1 NP_001128516.1
          NM_002752.4 NP_002743.3
          NM_139068.2 NP_620707.1
          NM_139069.2 NP_620708.1
          NM_139070.2 NP_620709.1
          XM_005265940.4 XP_005265997.1
          XM_006714891.2 XP_006714954.1
          XM_017009638.1 XP_016865127.1
          XM_017009639.1 XP_016865128.1
          XM_017009640.1 XP_016865129.1
          XM_017009641.1 XP_016865130.1
          XM_017009642.1 XP_016865131.1
          XM_017009643.1 XP_016865132.1
          AK289638.1
          DB451562.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv1683e212 Chr.5:180267143 - 180299506 on Build GRCh38 Gain MAPK9
          nsv1033706 Chr.5:179975769 - 180379356 on Build GRCh38 Gain RASGEF1C MAPK9 RNF130 MIR340 GFPT2
          esv2759398 Chr.5:180107477 - 180302944 on Build GRCh38 Gain RASGEF1C MAPK9 GFPT2

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          release of cytochrome c from mitochondria
          positive regulation of protein phosphorylation
          protein phosphorylation
          protein targeting to mitochondrion
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          response to stress
          JNK cascade
          JUN phosphorylation
          central nervous system development
          response to mechanical stimulus
          response to toxic substance
          positive regulation of gene expression
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of cell morphogenesis involved in differentiation
          response to amine
          neuron projection development
          positive regulation of prostaglandin biosynthetic process
          regulation of protein ubiquitination
          positive regulation of prostaglandin secretion
          positive regulation of chemokine production
          cellular response to UV
          Fc-epsilon receptor signaling pathway
          response to drug
          regulation of circadian rhythm
          positive regulation of nitric oxide biosynthetic process
          positive regulation of transcription, DNA-templated
          regulation of JNK cascade
          response to cadmium ion
          rhythmic process
          regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of nitric-oxide synthase biosynthetic process
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to growth factor stimulus
          positive regulation of podosome assembly
          positive regulation of apoptotic signaling pathway
          JUN kinase activity
          protein binding
          ATP binding
          transcription factor binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          mitogen-activated protein kinase kinase kinase binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline